Di-tert-butyl iminodicarboxylate, 97%
Di-tert-butyl iminodicarboxylate, 97%
Di-tert-butyl iminodicarboxylate, 97%
Thermo Scientific Chemicals

Di-tert-butyl iminodicarboxylate, 97%

CAS: 51779-32-9 | C10H19NO4 | 217.27 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass Bottle
Catalog number 369660050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS51779-32-9
IUPAC Nametert-butyl N-[(tert-butoxy)carbonyl]carbamate
Molecular FormulaC10H19NO4
InChI KeyXCAQIUOFDMREBA-UHFFFAOYSA-N
SMILESCC(C)(C)OC(=O)NC(=O)OC(C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Crystalline powder
Appearance (Color)White to beige
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only