PMSF Protease Inhibitor - FAQs
View additional product information for PMSF Protease Inhibitor - FAQs (36978)
1 product FAQs found
How should I reconstitute and store PMSF Protease Inhibitor (Cat. No. 36978)?
We recommend dissolving PMSF Protease Inhibitor (Cat. No. 36978) in isopropanol at 1.74 mg/mL (10 mM) and storing in aliquots at -20 degrees C.
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.