N-BOC-N,2-dimetilalanina, 98+ %, Thermo Scientific Chemicals
N-BOC-N,2-dimetilalanina, 98+ %, Thermo Scientific Chemicals
N-BOC-N,2-dimetilalanina, 98+ %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N-BOC-N,2-dimetilalanina, 98+ %, Thermo Scientific Chemicals

CAS: 146000-39-7 | C10H19NO4 | 217.27 g/mol
Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
1 g
5 g
Envase:
Frasco de vidrio
Número de catálogo 370950010
Precio (EUR)
71,20
Each
Cantidad:
1 g
Envase:
Frasco de vidrio
Pedido a granel o personalizado
Precio (EUR)
71,20
Each
Identificadores químicos
CAS146000-39-7
IUPAC Name2-{[(tert-butoxy)carbonyl](methyl)amino}-2-methylpropanoic acid
Molecular FormulaC10H19NO4
InChI KeyKPDLITGXUYMJEC-UHFFFAOYSA-N
SMILESCN(C(=O)OC(C)(C)C)C(C)(C)C(O)=O
Ver más
EspecificacionesSpecification SheetHoja de especificaciones
Appearance (Color)White
Infrared spectrumConforms
Melting point153°C to 157°C
Appearance (Form)Crystalline powder
Titration with NaOH>=98.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only