Matrix™ 2D Barcoded Open-Top Storage Tubes
Matrix™ 2D Barcoded Open-Top Storage Tubes
Matrix™ 2D Barcoded Open-Top Storage Tubes
Matrix™ 2D Barcoded Open-Top Storage Tubes
Matrix™ 2D Barcoded Open-Top Storage Tubes

Matrix™ 2D Barcoded Open-Top Storage Tubes

Green features
Store and track sample material in these 2D barcoded clear polypropylene open-top storage tubes.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)SterilityWell ShapeFit for Purpose
3712-111.4 mLSterileFlatSterile and PCR Ready
3719-111.4 mLNon-sterileFlatPCR Ready
3725TS0.5 mLSterileV-bottomSterile and PCR Ready
3715-111.4 mLNon-sterileFlatPCR Ready
37101.4 mLNon-sterileFlatPCR Ready
3730-110.75 mLNon-sterileV-bottomPCR Ready
3731-110.75 mLNon-sterileV-bottomPCR Ready
37111.4 mLNon-sterileFlatPCR Ready
37121.4 mLSterileFlatSterile and PCR Ready
37300.75 mLNon-sterileV-bottomSterile and PCR Ready
37320.75 mLSterileV-bottomSterile and PCR Ready
37340.5 mLNon-sterileV-bottomPCR Ready
3735500 μLSterileV-bottomSterile and PCR Ready
3750500 μLNon-sterileV-bottomPCR Ready
37901.4 mLNon-sterileV-bottomPCR Ready
37911.4 mLNon-sterileV-bottomPCR Ready
37921.4 mLSterileV-bottomSterile and PCR Ready
Catalog number 3712-11
Price (USD)
739.94
Online exclusive
822.15
Save 82.21
Each
Capacity (Metric):
1.4 mL
Sterility:
Sterile
Well Shape:
Flat
Fit for Purpose:
Sterile and PCR Ready
Price (USD)
739.94
Online exclusive
822.15
Save 82.21
Each

Store and track samples in Thermo Scientific™ Matrix™ 2D Barcoded Clear Polypropylene Open-Top Storage Tubes. Laser etching produces a permanent, high-contrast, 2D barcode offering maximum reliability and traceability of compound, biological and genomic samples. Offered in three sizes, 0.5mL, 0.75mL, and 1.4mL, these tubes can be sealed with Thermo Scientific SepraSeal septum caps and stored down to -80°C. Item number 3792 is ACT Labeled by My Green Lab to enable data-based sustainable product purchasing decisions.

Secure Tracking:

  • A unique 2D barcode is laser-etched onto the base of every polypropylene open-top tube to securely identify and track samples.
  • 2D barcodes are resistant to 100% DMSO, ideal for compound processing and storage.
  • Non-proprietary 2D codes can be scanned in less than a second in VisionMate High Speed for fast decoding and sample tracking.

Robust Storage Rack Design:

  • Matrix 2D barcoded open-top storage tubes come racked in 96-format in a proprietary specially designed, stackable, ANSI footprint.
  • Latch racks save precious space in your freezer, refrigerator, compound store or on the bench top.
  • The latch rack design enables manual multichannel pipette access to 2D tubes and eliminates contamination risk with lid design that does not contact the benchtop.
  • The latch rack lid can be lifted robotically for 2D tube access with automated liquid handling systems and high throughput applications.

Stringent Quality Control:

  • Every 2D barcoded storage tube is scanned to guarantee readability.
  • Each code is checked against a full database of all previously assigned 2D codes to ensure zero duplicates across the entire line of Matrix 2D barcoded storage tubes.
  • Every storage tube is leak tested to ensure the integrity and security of samples.
  • Matrix storage tubes are manufactured from medical grade virgin polypropylene
  • Tubes are validated to be free of DNA, DNAse, RNAse, PCR Inhibitors, endotoxins and cytotoxins

Storage Tracking Solutions for Any Laboratory:

  • Tubes are available with a range of sealing options including solid and pre-slit Duraseals and Sepraseals to meet your individual storage requirements.
  • Custom 2D coding options are available to meet your laboratory's individual database or tracking requirements.
  • 2D storage tubes are compatible with Thermo Scientific freezer racks that allow optimal utilization of storage space.
  • Item 3710 participates in My Green Lab's ACT Label program for Accountability, Consistency and Transparency (ACT) around evaluating product sustainability

Recommended for:

Individual sample retrieval, instant sample identification, secure identification of archived samples, secure shipping of samples, storage of samples and tracking of samples.

Recycling Information:

Matrix tubes, tube racks and covers are made from polypropylene copolymer and are recyclable in communities where polypropylene is recycled. Please recycle only clean, dry components that have been decontaminated of any hazardous materials or residues. The orange nylon rack latches must be removed from the tube rack base and discarded before recycling the rack base. Tip: Orange latches can be removed with relative ease with the aid of pliers.

Matrix tubes are delivered in high graphic paper cartons. Please recycle the paper cartons and product inserts.

Specifications
Capacity (English)0.0473 fl oz.
Green FeaturesACT Ecolabel (Gold)
Packaging10 Latch Racks of 96 Tubes
Well ShapeFlat
Bottom ShapeFlat
Capacity (Metric)1.4 mL
Fit for PurposeSterile and PCR Ready
MaterialPolypropylene
Product TypeStorage Tube (Open Top)
SterilitySterile
Unit SizeEach

Frequently asked questions (FAQs)

Can I store the Thermo Scientific 2D barcoded storage tubes in standard 10x10 cryoboxes?

Yes, the Thermo Scientific 2D barcoded storage tubes can be stored in standard cryoboxes. However, we do not carry 2D barcode readers capable of reading a full box of 2D barcodes in this storage format.

How do you prevent 2D barcode duplication during manufacturing and ensure quality control of the Thermo Scientific 2D barcoded storage tubes?

The 2D barcode laser etching software includes algorithms that prevent the barcode generator from ever counting backwards, subsequently preventing the re-etching of any previously generated barcode. To verify the above procedure, several quality control checks are performed on the 2D barcodes including:
- Checking for light transmission through the barcode as light transmission will reduce readability
- Inspecting the barcode under magnification
- Reading the barcode on a full rack 2D barcode reader
- Checking the barcode against a database of previously etched 2D barcodes
As the above quality control steps are completed, defective storage tubes are discarded, replaced, and re-checked, ensuring that the 2D barcodes are 100% readable and have not been previously used. This quality control process is the reason why we cannot guarantee sequential 2D barcode printing within a single rack of 2D barcoded products.

Can the barcodes on the Thermo Scientific 2D barcoded storage tubes be damaged by chemicals or by exposure to extreme temperatures?

The Thermo Scientific 2D barcoded storage tubes have been subjected in-house to consecutive liquid nitrogen freeze-thaws followed by submersion in boiling water. These tests did not cause any damage to the 2D barcode.
The 2D barcodes are made of the same material as the storage tubes and are resistant to standard lab chemicals.
The 2D barcode has a built-in ECC200 error barcode correction, which ensures that a damaged barcode is still readable as long as less than 25% of the barcode is missing.