Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • EphA4 Antibodies

          Invitrogen

          EphA4 Monoclonal Antibody (4C8H5)

          Advanced Verification
          17 Published Figures
          37 References
          View all (25) EphA4 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite EphA4 Monoclonal Antibody (4C8H5)

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Published Figures (17)
          • Advanced Verification (1)
          EphA4 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          EphA4 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 18

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          EphA4 Antibody (37-1600) in ICC/IF

          Molecular and cellular biology 2013 - Published figure using EphA4 monoclonal antibody (Product # 37-1600) in Immunocytochemistry {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          EphA4 Antibody in Immunocytochemistry (ICC/IF)
          EphA4 Antibody in Immunohistochemistry (IHC)
          EphA4 Antibody in Western Blot (WB)
          EphA4 Antibody in Western Blot (WB)
          EphA4 Antibody in Western Blot (WB)
          EphA4 Antibody in Western Blot, Immunoprecipitation (WB, IP)
          EphA4 Antibody in Western Blot (WB)
          EphA4 Antibody in Western Blot (WB)
          EphA4 Antibody in Western Blot (WB)
          EphA4 Antibody in Western Blot (WB)
          EphA4 Antibody in Western Blot (WB)
          EphA4 Antibody in Western Blot (WB)
          EphA4 Antibody in Western Blot (WB)
          EphA4 Antibody in Western Blot (WB)
          EphA4 Antibody in Western Blot (WB)
          EphA4 Antibody in Western Blot (WB)
          EphA4 Antibody in Western Blot (WB)
          EphA4 Antibody

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          EphA4 Monoclonal Antibody (4C8H5)

          Product Details

          37-1600

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1-3 µg/mL
          View 28 publications 28 publications

          Immunohistochemistry (IHC)

          3-10 µg/mL
          View 3 publications 3 publications

          Immunocytochemistry (ICC/IF)

          -
          View 2 publications 2 publications

          ELISA (ELISA)

          Assay-dependent
          -

          Immunoprecipitation (IP)

          5 µg
          View 3 publications 3 publications

          Inhibition Assays (Inhibition)

          -
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Chicken, Human, Mouse

          Published species

          Human, Mouse

          Host/Isotype

          Mouse / IgG1, kappa

          Class

          Monoclonal

          Type

          Antibody

          Clone

          4C8H5

          Immunogen

          A synthetic peptide derived from the C-terminus human EphA4 receptor protein.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.5 mg/mL

          Amount

          100 µg

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          0.1% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2533301

          Product Specific Information

          This antibody is specific for the endogenous EphA4 receptor protein. No cross-reactivity with EphA5 or EphA7 has been detected. On western blots, this antibody identifies a band at ~110-120 kDa. Reactivity has been confirmed with EphA4-transfected 293T cells and mouse brain lysates by western blot and immunoprecipitation and on transfected 293 cells and mouse hippocampus by immunohistochemistry.

          Target Information

          EPHA4 belongs to the ephrin receptor subfamily of the protein tyrosine kinase family. EPH and EPH-related receptors have been implicated in mediated developmental events, particularly in the nervous system. Receptors in the EPH subfamily typically have a single kinase domain and an extracellular region containing a Cys-rich domain and 2 fibronectin type III repeats. The ephrin receptors are divided into 2 groups based on the similarity of their extracellular domain sequences and their affinities for binding ephrin-A and ephrin-B ligands. EphA4 encodes a protein that binds ephrin-A ligands.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: Cek8; EK8; epa4; EPH-like kinase 8; Eph-related receptor protein tyrosine kinase; receptor protein-tyrosine kinase HEK8; TYRO1 protein tyrosine kinase; tyrosine-protein kinase receptor MPK-3; tyrosine-protein kinase receptor SEK; tyrosine-protein kinase receptor SEK-1; tyrosine-protein kinase TYRO1; unnamed protein product

          View more View less

          Gene Aliases: 2900005C20Rik; Cek8; EK8; HEK8; rb; SEK; Sek1; TYRO1

          View more View less

          UniProt ID: (Chicken) Q07496, (Human) P54764, (Mouse) Q03137

          View more View less

          Entrez Gene ID: (Chicken) 395559, (Human) 2043, (Mouse) 13838

          View more View less

          Function(s)
          protein kinase activity transmembrane receptor protein tyrosine kinase activity GPI-linked ephrin receptor activity transmembrane-ephrin receptor activity ATP binding PH domain binding identical protein binding DH domain binding protein tyrosine kinase binding beta-amyloid binding protein tyrosine kinase activity ephrin receptor activity protein binding kinase activity ephrin receptor binding transmembrane signal receptor
          Process(es)
          somitogenesis cell adhesion transmembrane receptor protein tyrosine kinase signaling pathway multicellular organism development axon guidance adult walking behavior motor neuron axon guidance peptidyl-tyrosine phosphorylation corticospinal tract morphogenesis positive regulation of kinase activity adherens junction organization segmentation regulation of GTPase activity positive regulation of JUN kinase activity protein autophosphorylation ephrin receptor signaling pathway negative regulation of axon regeneration regulation of astrocyte differentiation regulation of axonogenesis protein stabilization regulation of dendritic spine morphogenesis positive regulation of protein tyrosine kinase activity nephric duct morphogenesis cochlea development fasciculation of sensory neuron axon fasciculation of motor neuron axon synapse pruning negative regulation of long-term synaptic potentiation positive regulation of beta-amyloid formation positive regulation of aspartic-type endopeptidase activity involved in amyloid precursor protein catabolic process negative regulation of proteolysis involved in cellular protein catabolic process cellular response to beta-amyloid regulation of modification of synaptic structure positive regulation of Rho guanyl-nucleotide exchange factor activity negative regulation of cell adhesion positive regulation of cell proliferation glial cell migration negative regulation of epithelial to mesenchymal transition negative regulation of neuron projection development negative regulation of translation positive regulation of cell migration negative regulation of cell migration positive regulation of cell adhesion positive regulation of JNK cascade positive regulation of dendrite morphogenesis negative regulation of ERK1 and ERK2 cascade neuron projection guidance neuron projection fasciculation negative regulation of cellular response to hypoxia positive regulation of intracellular signal transduction positive regulation of amyloid precursor protein catabolic process positive regulation of leukocyte adhesion to arterial endothelial cell regulation of synapse pruning deadenylation-dependent decapping of nuclear-transcribed mRNA central nervous system projection neuron axonogenesis negative regulation of neuron apoptotic process negative regulation of translational initiation animal organ development regulation of developmental process synapse organization innervation membraneless organelle assembly cellular response to oxygen-containing compound
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5db85fbf84-frpqk:80/100.66.72.150:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch