Nunc™ 1.8mL Externally-Threaded Universal Tubes with 1D (linear) code on the side and 2D code on tube bottom, not racked - FAQs
View additional product information for Nunc™ 1.8mL Externally-Threaded Universal Tubes and Racks - FAQs (374501, 374502, 374500TS, 374501-BR, 374500-BR, 374502NOV, 374503)
1 product FAQs found
What is the Thermo Fisher alternative to Corning® 2 mL Internal Threaded Polypropylene Cryogenic Vial with 1D and 2D Bar codes, Product Number 8670?
The following alternatives for the Corning Cryogenic Vial, Product Number 8670:
- Nunc 2.0 mL Internally-Threaded Universal Tubes (Cat. No. 374510, 374510-BR)
- Nunc 2.0 mL Internally-Threaded Universal Tubes (Cat. No. 374512). Please note that this product is FDA registered and CE-marked.
- Nunc 1.8mL Externally-Threaded Universal Tubes and Racks (Cat. No. 374502, 374502NOV
- Matrix ScrewTop Tri-coded Tubes (Cat. No. 3740-WP1D)