Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 4
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Both IHC and WB applications require the following deglycosylation protocols.
Immunohistochemistry
1. Prepare the following stock solutions:
a. 200 mM NaIO4 (8.556 g/200 mL water)
b. Sodium-Acetate buffer, pH 4.5: Dissolve NaCl (final concentration: 0.33M) into 0.1M glacial acetic acid and adjust to pH 4.5 using 1 N NaOH.
c. Sodium-thiosulfate solution: Dissolve Na2S2O3 (0.133M), NaI (0.033M), and NaHCO3 (0.033M) in water and adjust to pH 7.6 using 1M HCl. Keep solution at 4°C.
2. After deparaffinization, rehydration, and peroxidase quenching steps, mix a 1:1 solution of 'a' and 'b' (in step 1), add to each slide, and incubate overnight at 4°C.
3. Treat tissue with solution 'c' for 30 minutes at 4°C.
4. Proceed with normal immunostaining protocol (blocking and antibody incubation steps).
Western blotting
1. Treat transblotted membrane with 10 mM NaIO4 in 0.1M sodium acetate/5 mM EDTA, pH 5.5 overnight at 4°C in the dark.
2. After washing off the solution, incubate the membrane in a solution containing 30 mM Na2S2O3, 7.5 mM NaI, and 7.5 mM NaHCO3, pH 7.2 for 1 hour at 4°C in the dark.
3. Proceed with normal Western blot protocol (wash and blocking steps).
Mucins (MUCs) are heavily glycosylated glycoproteins that are synthesized either as membrane-bound or secreted proteins. Mucins are noteworthy because they have an aberrant expression profile in various malignancies. Mucins normally play a protective role for epithelial tissues and are expressed in the gastrointestinal, respiratory, and genito-urinary tracts. Mucins may also be involved in the renewal and differentiation of the epithelium, and the modulation of cell adhesion and cell signaling. MUC5B is thought to play a vital role in maintaining oral health, and may also be involved in respiratory and oral disease.2 MUC5B expression has been studied in gastric carcinoma as well as chronic obstructive pulmonary disease.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: cervical mucin MUC5B; high molecular weight mucin; high molecular weight salivary mucin MG1; MUC5B; mucin 5, subtype B, tracheobronchial; mucin MG1; sublingual gland mucin; unnamed protein product
Gene Aliases: MG1; MUC-5B; MUC5; MUC9
UniProt ID: (Human) Q9HC84
Entrez Gene ID: (Human) 727897
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support