D(+)-Glucose, anhydrous, specificied according to the requirements of Ph.Eur., USP, BP
D(+)-Glucose, anhydrous, specificied according to the requirements of Ph.Eur., USP, BP
D(+)-Glucose, anhydrous, specificied according to the requirements of Ph.Eur., USP, BP
Thermo Scientific Chemicals

D(+)-Glucose, anhydrous, specificied according to the requirements of Ph.Eur., USP, BP

CAS: 50-99-7 | C6H12O6 | 180.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
5 kg
25 kg
Catalog number 388190050
Price (USD)
150.00
Each
Quantity:
5 kg
Request bulk or custom format
Price (USD)
150.00
Each
Chemical Identifiers
CAS50-99-7
IUPAC Name6-(hydroxymethyl)oxane-2,3,4,5-tetrol
Molecular FormulaC6H12O6
InChI KeyWQZGKKKJIJFFOK-UHFFFAOYNA-N
SMILESOCC1OC(O)C(O)C(O)C1O
View more
SpecificationsSpecification SheetSpecification Sheet
Soluble starch, sulfite=<15 ppm (EP)
Sulfated ash=<0.1 % (1 g, 600°C)
Arsenic (As)=<1 µg/g
Appearance (Form)Crystalline powder
Specific optical rotation+52.5° to +53.3° (20°C, 589 nm) (C=10,H2O/dilute NH3)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only