Triethylamine Acetate (TEAA) 2.0 M - FAQs
Triethylamine Acetate (TEAA) 2.0 M - FAQs
View additional product information for Triethylamine Acetate (TEAA) 2.0 M - FAQs (400613)
1 product FAQs found
Do you know of any alternatives for Triethylamine Acetate (TEAA) 2.0 M (Cat. No. 400613)?
We do not offer any alternatives for Triethylamine Acetate (TEAA) 2.0 M.