GeneScan™ 1000 ROX™ dye Size Standard - FAQs
GeneScan™ 1000 ROX™ dye Size Standard - FAQs
View additional product information for GeneScan™ 1000 ROX™ dye Size Standard - FAQs (401098)
1 product FAQs found
If my fragments are greater than 600 bp, can I use the GeneScan 1200 LIZ Dye, GeneScan 1000 ROX Dye, or GeneScan 2500 ROX Dye Size Standards?
For fragments that are greater than 600 bp, the GeneScan 1200 LIZ Dye Size Standard is recommended. The GeneScan 1000 ROX Dye, or GeneScan 2500 ROX Dye Size Standards are intended for non-denaturing applications only.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Applications Support Center.