Aluminon, ACS reagent - FAQs
Aluminon, ACS reagent - FAQs
View additional product information for Aluminon, ACS reagent - FAQs (401500250, 401501000)
1 product FAQs found
What is the solubility of Aluminon?
Aluminon is soluble in water at a concentration of ~800 g/L at 20°C. It is slightly soluble in ethanol and insoluble in petroleum ether.