beta-Cyclodextrin, 98%
beta-Cyclodextrin, 98%
beta-Cyclodextrin, 98%
Thermo Scientific Chemicals

beta-Cyclodextrin, 98%

CAS: 7585-39-9 | C42H70O35 | 1134.99 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number 406001000
Price (EUR)
63,65
Online Exclusive
70,40
Save 6,75 (10%)
Each
Quantity:
100 g
Request bulk or custom format
Price (EUR)
63,65
Online Exclusive
70,40
Save 6,75 (10%)
Each
Chemical Identifiers
CAS7585-39-9
IUPAC Name5,10,15,20,25,30,35-heptakis(hydroxymethyl)-2,4,7,9,12,14,17,19,22,24,27,29,32,34-tetradecaoxaoctacyclo[31.2.2.2³,⁶.2⁸,¹¹.2¹³,¹⁶.2¹⁸,²¹.2²³,²⁶.2²⁸,³¹]nonatetracontane-36,37,38,39,40,41,42,43,44,45,46,47,48,49-tetradecol
Molecular FormulaC42H70O35
InChI KeyWHGYBXFWUBPSRW-UHFFFAOYNA-N
SMILESOCC1OC2OC3C(CO)OC(OC4C(CO)OC(OC5C(CO)OC(OC6C(CO)OC(OC7C(CO)OC(OC8C(CO)OC(OC1C(O)C2O)C(O)C8O)C(O)C7O)C(O)C6O)C(O)C5O)C(O)C4O)C(O)C3O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Specific optical rotation+159° to +165° (20°C, 589 nm) (c=1.5, water)
Appearance (Color)White
HPLC>=97.5 %
Specific optical rotationon dry substance
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only