Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Alpha-Smooth Muscle Actin Antibodies

          Invitrogen

          Alpha-Smooth Muscle Actin Monoclonal Antibody (1A4), eFluor™ 570, eBioscience™

          28 Published Figures
          36 References
          View all (78) Alpha-Smooth Muscle Actin antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite Alpha-Smooth Muscle Actin Monoclonal Antibody (1A4), eFluor™ 570, eBioscience™

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (2)
          • Published Figures (28)
          Alpha-Smooth Muscle Actin Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          Alpha-Smooth Muscle Actin Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.

          FIGURE: 1 / 30

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Alpha-Smooth Muscle Actin Antibody (41-9760-80) in IHC (P)

          Immunohistochemistry of formalin-fixed paraffin embedded human infiltrating ductal carcinoma tissue using 1 µg/mL Mouse IgG2a, K Isotype Control eFluor® 570 (left) or 1 µg/mL Anti-Alpha-Smooth Muscle Actin eFluor® 570 (right). Nuclei are stained with DAPI. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Alpha-Smooth Muscle Actin Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Alpha-Smooth Muscle Actin Antibody in Multiplex Immunohistochemistry (mIHC)
          Alpha-Smooth Muscle Actin Antibody in Immunohistochemistry (IHC)
          Alpha-Smooth Muscle Actin Antibody in Immunocytochemistry (ICC/IF)
          Alpha-Smooth Muscle Actin Antibody in Immunocytochemistry (ICC/IF)
          Alpha-Smooth Muscle Actin Antibody in Immunocytochemistry (ICC/IF)
          Alpha-Smooth Muscle Actin Antibody in Immunocytochemistry (ICC/IF)
          Alpha-Smooth Muscle Actin Antibody in Immunocytochemistry (ICC/IF)
          Alpha-Smooth Muscle Actin Antibody in Immunocytochemistry (ICC/IF)
          Alpha-Smooth Muscle Actin Antibody in Immunocytochemistry (ICC/IF)
          Alpha-Smooth Muscle Actin Antibody in Immunocytochemistry (ICC/IF)
          Alpha-Smooth Muscle Actin Antibody in Immunocytochemistry (ICC/IF)
          Alpha-Smooth Muscle Actin Antibody in Immunocytochemistry (ICC/IF)
          Alpha-Smooth Muscle Actin Antibody in Immunocytochemistry (ICC/IF)
          Alpha-Smooth Muscle Actin Antibody in Immunocytochemistry (ICC/IF)
          Alpha-Smooth Muscle Actin Antibody in Immunocytochemistry (ICC/IF)
          Alpha-Smooth Muscle Actin Antibody in Immunohistochemistry, Immunocytochemistry (IHC, ICC/IF)
          Alpha-Smooth Muscle Actin Antibody in Immunohistochemistry (IHC)
          Alpha-Smooth Muscle Actin Antibody in Immunohistochemistry (IHC)
          Alpha-Smooth Muscle Actin Antibody in Immunohistochemistry (IHC)
          Alpha-Smooth Muscle Actin Antibody in Immunohistochemistry (IHC)
          Alpha-Smooth Muscle Actin Antibody in Immunohistochemistry (IHC)
          Alpha-Smooth Muscle Actin Antibody in Immunohistochemistry (IHC)
          Alpha-Smooth Muscle Actin Antibody in Immunohistochemistry (IHC)
          Alpha-Smooth Muscle Actin Antibody in Immunohistochemistry (IHC)
          Alpha-Smooth Muscle Actin Antibody in Immunohistochemistry (IHC)
          Alpha-Smooth Muscle Actin Antibody in Immunohistochemistry (IHC)
          Alpha-Smooth Muscle Actin Antibody in Western Blot (WB)
          Alpha-Smooth Muscle Actin Antibody in Western Blot (WB)
          Alpha-Smooth Muscle Actin Antibody in Western Blot (WB)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Product Details

          41-9760-80

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          -
          View 2 publications 2 publications

          Immunohistochemistry (IHC)

          1:100
          View 19 publications 19 publications

          Multiplex Immunohistochemistry (mIHC)

          1 µg/mL
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          1 µg/mL
          View 1 publication 1 publication

          Immunohistochemistry (Frozen) (IHC (F))

          1 µg/mL
          -

          Immunocytochemistry (ICC/IF)

          Assay-Dependent
          View 10 publications 10 publications

          Flow Cytometry (Flow)

          -
          View 4 publications 4 publications
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Published species

          Human, Mouse

          Host/Isotype

          Mouse / IgG2a, kappa

          Class

          Monoclonal

          Type

          Antibody

          Clone

          1A4

          Conjugate

          eFluor™ 570 eFluor™ 570 eFluor™ 570
          • Unconjugated
          • Alexa Fluor 420 
          • Alexa Fluor 488 
          • Alexa Fluor 514 
          • Alexa Fluor 700 
          • Alexa Fluor Plus 488 
          • Alexa Fluor Plus 594 
          • Alexa Fluor Plus 750
          • eFluor 660
          • Request custom conjugation

          Excitation/Emission Max

          556/569 nm View spectra spectra

          Form

          Liquid

          Concentration

          0.2 mg/mL

          Amount

          25 µg

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.2

          Contains

          0.09% sodium azide

          Storage conditions

          4°C, store in dark, DO NOT FREEZE!

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2573630

          Product Specific Information

          Description: The monoclonal antibody 1A4 recognizes human, mouse, and rat alpha-smooth muscle actin. Alpha-smooth muscle actin is a 42 kDa protein and is a major component of the cytoskeletal structural network. This 1A4 antibody is specific for the alpha form of muscle actin only, which is expressed by smooth muscle cells of blood vessels, myofibroblasts, and myoepithelial cells. It is also expressed in the parenchyma and stroma of various tissues. Alpha-smooth muscle actin is useful in the identification of leiomyomas, leiomycosarcomas and pleomorphic adenomas and is used as a prognostic marker for basal cell carcinoma.

          Applications Reported: This 1A4 antibody has been reported for use in immunohistochemical staining of frozen tissue sections, immunohistochemical staining of formalin-fixed paraffin embedded tissue sections, microscopy, and immunocytochemistry.

          Applications Tested: This 1A4 antibody has been tested by immunohistochemistry of formalin-fixed paraffin embedded human tissue using low pH antigen retrieval and can be used at less than or equal to 1 µg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.

          Filter Recommendation: When using this eFluor™ 570 antibody conjugate, we recommend a filter that will capture the 570 emission wavelength (for example, Excitation 545/25, 565LP, Emission 605/70). A standard Alexa Fluor™ 555 or TRITC filter is acceptable.

          Excitation: 555 nm; Emission: 570 nm

          Target Information

          Smooth Muscle Actin belongs to the actin family of proteins, which are highly conserved proteins that play a role in cell motility, structure and integrity. Alpha, beta and gamma actin isoforms have been identified, with alpha actin being a major constituent of the contractile apparatus, while beta and gamma actins are involved in the regulation of cell motility. In particular, smooth muscle actin is an alpha actin that is found in skeletal muscle. Actin exists as a ubiquitous protein involved with filament formation that make up large portions of the cytoskeleton. Actin filaments interact with myosin to assist in muscle contraction as well as aiding in cell motility and cytokinesis. Smooth muscle actin is found on smooth muscle vessel walls, gut wall, myometrium, myoepithelial cells in breast and salivary glands. Defects in the smooth muscle actin gene cause aortic aneurysm familial thoracic type 6. Actin isoforms differ slightly in their N-terminus and the sequences of each are perfectly conserved in higher vertebrates. Alpha-smooth muscle actin is abundant in vascular and visceral smooth muscle cells. In addition, it has also been shown that smooth muscle actin appear in stress fibers of fibroblastic cells during pathological situations involving contractile phenomena such as wound healing and fibrocontractive diseases. Multiple alternatively spliced variants of smooth muscle actin have been identified.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: ACTA2; Actin Alpha 2 Smooth Muscle Aorta; Actin Vascular Smooth Muscle; actin, alpha 2, smooth muscle, aorta; actin, alpha, vascular smooth muscle; Alpha Cardiac Actin; alpha SMA; alpha-actin; alpha-actin-2; alpha-cardiac actin; alpha-smooth muscle actin; Aortic Smooth Muscle; asma; aSmooth Muscle Actin; cell growth-inhibiting gene 46 protein; GIG46; Growth Inhibiting Gene 46; smooth muscle; smooth muscle alpha-actin; unnamed protein product

          View more View less

          Gene Aliases: 0610041G09Rik; a-SMA; ACTSA; Actvs; alphaSMA; SMAalpha; SMalphaA; SMDYS

          View more View less

          UniProt ID: (Human) P62736, (Rat) P62738, (Mouse) P62737

          View more View less

          Entrez Gene ID: (Human) 59, (Rat) 81633, (Mouse) 11475

          View more View less

          Function(s)
          structural constituent of cytoskeleton protein binding ATPase activity protein kinase binding
          Process(es)
          muscle contraction cytoskeleton organization regulation of blood pressure response to virus positive regulation of gene expression vascular smooth muscle contraction positive regulation of hepatic stellate cell migration positive regulation of hepatic stellate cell contraction positive regulation of ERK1 and ERK2 cascade cellular response to transforming growth factor beta stimulus juxtaglomerular apparatus development glomerular mesangial cell development mesenchyme migration positive regulation of hepatic stellate cell activation
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5db85fbf84-q86qf:80/100.66.76.64:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch