Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • NOTCH1 Antibodies

          Invitrogen

          NOTCH1 Monoclonal Antibody (mN1A)

          Advanced Verification
          2 References
          View all (59) NOTCH1 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite NOTCH1 Monoclonal Antibody (mN1A)

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          • Advanced Verification (2)
          NOTCH1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          NOTCH1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 3

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          NOTCH1 Antibody (41-3500) in WB

          Knockout of NOTCH1 was achieved by CRISPR-Cas9 genome editing using LentiArray™ Lentiviral sgRNA (Product # A32042, Assay ID CRISPR682588_LV) and LentiArray Cas9 Lentivirus (Product # A32064). Western blot analysis of NOTCH1 was performed by loading 50 µg of MDA-MB-231 wild type (Lane 1) and MDA-MB-231 NOTCH1 KO (Lane 2) whole cell extracts. The samples were electrophoresed using NuPAGE™ 3-8% Tris-Acetate Protein Gel (Product # EA0378BOX). Resolved proteins were then transferred onto a nitrocellulose membrane (Product # IB23001) by iBlot® ... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          NOTCH1 Antibody in Western Blot (WB)
          NOTCH1 Antibody
          NOTCH1 Antibody

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Product Details

          41-3500

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          Assay-dependent
          View 1 publication 1 publication

          Flow Cytometry (Flow)

          Assay-dependent
          -

          Immunoprecipitation (IP)

          -
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Bovine, Human, Mouse, Non-human primate, Rat

          Published species

          Human

          Host/Isotype

          Mouse / IgG1

          Class

          Monoclonal

          Type

          Antibody

          Clone

          mN1A

          Immunogen

          Recombinant protein derived from the intracellular domain of mouse Notch1

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.5 mg/mL

          Amount

          100 µg

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          0.1% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2533505

          Product Specific Information

          This antibody is specific for the Notch1 (neurogenic locus notch homolog protein1, Motch A, mT14, p300) protein. This antibody gives a band of ~65 kDa on Western blot analysis using Notch1-IC OP transfected cells. Western blot analysis using Jurkat cells expresses the target band at ~110-120 kDa, with an additional band occasionally observed at ~220kDa. This antibody does not cross-react with Notch 2,3, and 4 as assessed by Western blot analysis.

          Target Information

          The notch gene belongs to a family of epidermal growth factor (EGF) like homeotic genes, which encode transmembrane proteins with a variable number of cysteine-rich EGF-like repeats in the extracellular region. Four notch genes have been described in mammals: Notch1, Notch2, Notch3 and Notch4(Int-3), which have been implicated in the differentiation of the nervous system and other structures. The EGF-like proteins Delta and Serrate have been identified as ligands of Notch1. Mature Notch proteins are heterodimeric receptors derived from the cleavage of Notch pre-proteins into an extracellular subunit (NEC) containing multiple EGF-like repeats and a transmembrane subunit including intracellular region (Ntm).

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: Drosophila Notch homolog 1 (controlling the the ectodermal and neural cell fate in Drosophila); major type A protein; Motch A; mT14; neuro cell differentiation; notch 1; Notch gene homolog 1; Notch homolog 1, translocation-associated; Notch-like protein 1; NOTCH1; p300; RP23-306D20.12; RP23-306D20.12-001; TAN1; translocation-associated notch protein TAN-1; unnamed protein product

          View more View less

          Gene Aliases: 9930111A19Rik; AOS5; AOVD1; hN1; lin-12; Mis6; N1; NOTCH; TAN1

          View more View less

          UniProt ID: (Human) P46531, (Mouse) Q01705, (Rat) Q07008

          View more View less

          Entrez Gene ID: (Bovine) 767866, (Human) 4851, (Mouse) 18128, (Rat) 25496

          View more View less

          Function(s)
          chromatin binding transcription coactivator activity enzyme inhibitor activity transmembrane signaling receptor activity Notch binding calcium ion binding protein binding enzyme binding chromatin DNA binding signaling receptor activity identical protein binding transcription regulator activator activity extracellular matrix glycoprotein
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter luteolysis epithelial to mesenchymal transition heart looping positive regulation of neuroblast proliferation outflow tract morphogenesis endocardium development endocardium morphogenesis atrioventricular node development coronary vein morphogenesis aortic valve morphogenesis atrioventricular valve morphogenesis coronary sinus valve morphogenesis pulmonary valve morphogenesis mitral valve formation epithelial to mesenchymal transition involved in endocardial cushion formation endocardial cushion morphogenesis cardiac chamber formation cardiac ventricle morphogenesis cardiac atrium morphogenesis cardiac right atrium morphogenesis cardiac left ventricle morphogenesis ventricular trabecula myocardium morphogenesis growth involved in heart morphogenesis regulation of cardioblast proliferation cell migration involved in endocardial cushion formation negative regulation of extracellular matrix constituent secretion pericardium morphogenesis regulation of transcription, DNA-templated regulation of transcription from RNA polymerase II promoter protein import into nucleus immune response Notch signaling pathway positive regulation of transcription of Notch receptor target spermatogenesis determination of left/right symmetry axon guidance heart development positive regulation of cell proliferation negative regulation of cell proliferation gene expression negative regulation of cardiac muscle hypertrophy positive regulation of gene expression negative regulation of gene expression negative regulation of cardiac muscle cell apoptotic process positive regulation of epithelial to mesenchymal transition negative regulation of cell-substrate adhesion negative regulation of myotube differentiation mesenchymal cell development cell differentiation in spinal cord cell differentiation protein catabolic process negative regulation of ossification regulation of cell migration positive regulation of cell migration positive regulation of BMP signaling pathway negative regulation of BMP signaling pathway organ regeneration response to muramyl dipeptide response to lipopolysaccharide negative regulation of collagen biosynthetic process tube formation cellular response to vascular endothelial growth factor stimulus regulation of cell proliferation tissue regeneration negative regulation of catalytic activity positive regulation of viral genome replication positive regulation of endothelial cell differentiation negative regulation of myoblast differentiation negative regulation of neuron differentiation negative regulation of osteoblast differentiation positive regulation of glial cell differentiation negative regulation of transcription, DNA-templated positive regulation of transcription, DNA-templated positive regulation of transcription from RNA polymerase II promoter positive regulation of JAK-STAT cascade positive regulation of Ras protein signal transduction astrocyte differentiation oligodendrocyte differentiation positive regulation of astrocyte differentiation negative regulation of oligodendrocyte differentiation homeostasis of number of cells within a tissue negative regulation of neurogenesis regulation of developmental process positive regulation of smooth muscle cell differentiation cardiac muscle tissue morphogenesis cardiac muscle cell proliferation positive regulation of cardiac muscle cell proliferation negative regulation of glial cell proliferation cilium morphogenesis cardiac epithelial to mesenchymal transition negative regulation of cell adhesion molecule production cardiac muscle cell myoblast differentiation cardiac septum morphogenesis ventricular septum morphogenesis arterial endothelial cell differentiation venous endothelial cell differentiation cardiac vascular smooth muscle cell development endocardial cell differentiation vasculogenesis involved in coronary vascular morphogenesis coronary artery morphogenesis regulation of cell adhesion involved in heart morphogenesis heart trabecula morphogenesis positive regulation of cardiac epithelial to mesenchymal transition negative regulation of biomineral tissue development positive regulation of ERK1 and ERK2 cascade left/right axis specification cellular response to tumor cell cellular response to follicle-stimulating hormone stimulus cellular response to hypoxia distal tubule development collecting duct development glomerular mesangial cell development T-helper 17 type immune response negative regulation of cell migration involved in sprouting angiogenesis neuronal stem cell population maintenance interleukin-17-mediated signaling pathway chemical synaptic transmission, postsynaptic negative regulation of cold-induced thermogenesis regulation of extracellular matrix assembly positive regulation of apoptotic process involved in morphogenesis negative regulation of cell-cell adhesion mediated by cadherin negative regulation of cell proliferation involved in heart morphogenesis negative regulation of stem cell differentiation negative regulation of anoikis negative regulation of pro-B cell differentiation negative regulation of endothelial cell chemotaxis angiogenesis in utero embryonic development cell fate specification liver development morphogenesis of an epithelial sheet sprouting angiogenesis osteoblast fate commitment epithelial cell development inflammatory response to antigenic stimulus endocardial cushion development cardiac right ventricle formation humoral immune response compartment pattern specification axonogenesis foregut morphogenesis endoderm development epidermis development regulation of Notch signaling pathway auditory receptor cell fate commitment glial cell differentiation regulation of gene expression positive regulation of cell development regulation of somitogenesis neural tube development neuron differentiation keratinocyte differentiation lung development embryonic limb morphogenesis positive regulation of epithelial cell differentiation forebrain development hair follicle morphogenesis embryonic hindlimb morphogenesis epithelial cell differentiation involved in kidney development skeletal muscle cell differentiation regulation of apoptotic process positive regulation of apoptotic process negative regulation of programmed cell death cell fate commitment negative regulation of cell differentiation regulation of auditory receptor cell differentiation negative regulation of auditory receptor cell differentiation positive regulation of keratinocyte differentiation positive regulation of Notch signaling pathway negative regulation of calcium ion-dependent exocytosis negative regulation of photoreceptor cell differentiation somatic stem cell division developmental growth reproductive structure development neuron fate commitment branching morphogenesis of an epithelial tube artery morphogenesis venous blood vessel morphogenesis regulation of epithelial cell proliferation positive regulation of epithelial cell proliferation regulation of neurogenesis secretory columnal luminar epithelial cell differentiation involved in prostate glandular acinus development prostate gland epithelium morphogenesis regulation of epithelial cell proliferation involved in prostate gland development glomerulus vasculature development nephron tubule development negative regulation of canonical Wnt signaling pathway positive regulation of ephrin receptor signaling pathway apoptotic process involved in embryonic digit morphogenesis positive regulation of aorta morphogenesis regulation of cell proliferation involved in heart morphogenesis animal organ development Notch signaling involved in heart development
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-76c4fffd86-nv6df:80/100.66.78.131:80.
          git-commit: bdc067a1e9e92d056899a2dda64af631988aa8a1
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.54.0-Offline