1-Pyrrolidinecarbonyl chloride, 97%
1-Pyrrolidinecarbonyl chloride, 97%
1-Pyrrolidinecarbonyl chloride, 97%
Thermo Scientific Chemicals

1-Pyrrolidinecarbonyl chloride, 97%

CAS: 1192-63-8 | C5H8ClNO | 133.58 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass Bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 418650050
Price (USD)
63.65
Online exclusive
70.10
Save 6.45
Each
Quantity:
5 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)
63.65
Online exclusive
70.10
Save 6.45
Each
Chemical Identifiers
CAS1192-63-8
IUPAC Namepyrrolidine-1-carbonyl chloride
Molecular FormulaC5H8ClNO
InChI KeyXACWJIQLDLUFSR-UHFFFAOYSA-N
SMILESClC(=O)N1CCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Liquid
GC>=96.0 %
Appearance (Color)Clear colorless to light yellow
Refractive index1.4910 to 1.4970 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only