Sucrose octaacetate, 98%
Sucrose octaacetate, 98%
Sucrose octaacetate, 98%
Thermo Scientific Chemicals

Sucrose octaacetate, 98%

CAS: 126-14-7 | C28H38O19 | 678.59 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
5 kg
Packaging:
View all
Plastic Bottle
Catalog number 419790010
Price (USD)
102.65
Online exclusive
114.00
Save 11.35
Each
Quantity:
1 kg
Packaging:
Plastic Bottle
Request bulk or custom format
Price (USD)
102.65
Online exclusive
114.00
Save 11.35
Each
Chemical Identifiers
CAS126-14-7
IUPAC Name[(2R,3R,4S,5S)-3,4-bis(acetyloxy)-5-[(acetyloxy)methyl]-5-{[(2R,3R,4S,5R,6R)-3,4,5-tris(acetyloxy)-6-[(acetyloxy)methyl]oxan-2-yl]oxy}oxolan-2-yl]methyl acetate
Molecular FormulaC28H38O19
InChI KeyZIJKGAXBCRWEOL-SAXBRCJISA-N
SMILESCC(=O)OC[C@H]1O[C@@](COC(C)=O)(O[C@H]2O[C@H](COC(C)=O)[C@@H](OC(C)=O)[C@H](OC(C)=O)[C@H]2OC(C)=O)[C@@H](OC(C)=O)[C@@H]1OC(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
Appearance (Form)Powder
Titration excess NaOH97.5 to 102.5 % (on anhydrous substance)
Residue after ignition=<0.1 %
Melting point>=78°C
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only