Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Sigma-1 Receptor Antibodies
          Validated with Transparency. Independently tested through the YCharOS Open Science Initiative.

          Invitrogen

          Sigma-1 Receptor Polyclonal Antibody

          Advanced Verification
          27 Published Figures
          28 References
          View all (12) Sigma-1 Receptor antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite Sigma-1 Receptor Polyclonal Antibody

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (2)
          • Published Figures (27)
          • Advanced Verification (1)
          Sigma-1 Receptor Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          Sigma-1 Receptor Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 30

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Sigma-1 Receptor Antibody (42-3300) in WB

          Western blot of SIGMAR1 was performed by loading 30 µg of WT (lanes 1/3) and SIGMAR1 CRISPR KO (lanes 2/4) HEK293T or HeLa cell lysates in RIPA buffer onto a 5-16% gradient polyacrylamide gel. Proteins on the blots were visualized with Ponceau staining (below immunoblot). Proteins were transferred to nitrocellulose membrane and blocked in 5% milk for 1 hr. SIGMAR1 was detected at approximately 25 kDa (designated by the black arrow) using a SIGMAR1 polyclonal antibody (Product # 42-3300) at a dilution of 1:5,000 in 5% BSA in TBS with 0.1% T... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Sigma-1 Receptor Antibody in Western Blot (WB)
          Sigma-1 Receptor Antibody in Western Blot (WB)
          Sigma-1 Receptor Antibody in Immunocytochemistry (ICC/IF)
          Sigma-1 Receptor Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          Sigma-1 Receptor Antibody in Immunocytochemistry (ICC/IF)
          Sigma-1 Receptor Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          Sigma-1 Receptor Antibody in Immunocytochemistry (ICC/IF)
          Sigma-1 Receptor Antibody in Immunohistochemistry (IHC)
          Sigma-1 Receptor Antibody in Immunohistochemistry (IHC)
          Sigma-1 Receptor Antibody in Immunohistochemistry (IHC)
          Sigma-1 Receptor Antibody in Immunohistochemistry (IHC)
          Sigma-1 Receptor Antibody in Immunohistochemistry (IHC)
          Sigma-1 Receptor Antibody in Immunohistochemistry (IHC)
          Sigma-1 Receptor Antibody in Immunohistochemistry (IHC)
          Sigma-1 Receptor Antibody in Western Blot (WB)
          Sigma-1 Receptor Antibody in Western Blot (WB)
          Sigma-1 Receptor Antibody in Western Blot (WB)
          Sigma-1 Receptor Antibody in Western Blot (WB)
          Sigma-1 Receptor Antibody in Western Blot (WB)
          Sigma-1 Receptor Antibody in Western Blot (WB)
          Sigma-1 Receptor Antibody in Western Blot (WB)
          Sigma-1 Receptor Antibody in Western Blot (WB)
          Sigma-1 Receptor Antibody in Western Blot (WB)
          Sigma-1 Receptor Antibody in Western Blot (WB)
          Sigma-1 Receptor Antibody in Western Blot (WB)
          Sigma-1 Receptor Antibody in Western Blot (WB)
          Sigma-1 Receptor Antibody in Western Blot (WB)
          Sigma-1 Receptor Antibody in Western Blot (WB)
          Sigma-1 Receptor Antibody in Western Blot (WB)
          Sigma-1 Receptor Antibody

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Sigma-1 Receptor Polyclonal Antibody

          Product Details

          42-3300

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          2 µg/mL
          View 19 publications 19 publications

          Immunohistochemistry (IHC)

          -
          View 6 publications 6 publications

          Immunocytochemistry (ICC/IF)

          -
          View 2 publications 2 publications

          Functional Assay (Functional)

          -
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Mouse, Rat

          Published species

          Hamster, Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Synthetic peptide derived from the C-terminal region of the rat sigma-1 receptor, which differs from mouse and human by one conservative amino acid change and is predicted to be identical to dog, bovine, and ermine
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.25 mg/mL

          Amount

          100 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.4

          Contains

          0.1% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2533521

          Product Specific Information

          This antibody is specific for the sigma-1 receptor (sigma-1R, Oprs1 protein, opioid receptor sigma 1) protein. On western blots, it identifies the target band at ~25 kDa. Reactivity has been confirmed with rat brain, rat liver, and mouse brain homogenates. Based on amino acid sequence identity, reactivity with human, bovine, dog, and ermine is expected.

          Target Information

          This gene encodes a receptor protein that interacts with a variety of psychotomimetic drugs, including cocaine and amphetamines. The receptor is believed to play an important role in the cellular functions of various tissues associated with the endocrine, immune, and nervous systems. As indicated by its previous name, opioid receptor sigma 1 (OPRS1), the product of this gene was erroneously thought to function as an opioid receptor; it is now thought to be a non-opioid receptor. Alternative splicing of this gene results in transcript variants encoding distinct isoforms.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: AL024364; mSigmaR1; mSR-1A; mSR-1B; mSR-1C; mSR-1D; mSR-1E; opioid receptor, sigma 1; RP23 167I12.6; SIG 1R; sigma 1 receptor variant SR-1D; sigma 1-type opioid receptor; Sigma-1 Receptor; sigma1 receptor; sigma1-receptor; sigma1R; SR BP; unnamed protein product

          View more View less

          Gene Aliases: Oprs1; Sig1R; sigma1R

          View more View less

          UniProt ID: (Mouse) O55242, (Rat) Q9R0C9

          View more View less

          Entrez Gene ID: (Mouse) 18391, (Rat) 29336

          View more View less

          Function(s)
          opioid receptor activity protein binding mu-type opioid receptor binding signaling receptor activity identical protein binding
          Process(es)
          prostaglandin biosynthetic process nervous system development cyclooxygenase pathway opioid receptor signaling pathway regulation of neuron apoptotic process negative regulation of neuron apoptotic process negative regulation of nitric oxide biosynthetic process protein homotrimerization response to alcohol regulation of postsynapse assembly negative regulation of response to endoplasmic reticulum stress response to methamphetamine hydrochloride positive regulation of protein localization to nucleolus regulation of response to endoplasmic reticulum stress conditioned place preference
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5db85fbf84-lw26s:80/100.66.78.224:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch