cis-3-Azabicyclo[3.1.0]hexane-2-carboxylic acid, 95%
cis-3-Azabicyclo[3.1.0]hexane-2-carboxylic acid, 95%
cis-3-Azabicyclo[3.1.0]hexane-2-carboxylic acid, 95%
Thermo Scientific Chemicals

cis-3-Azabicyclo[3.1.0]hexane-2-carboxylic acid, 95%

CAS: 22255-16-9 | C6H9NO2 | 127.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Packaging:
Glass bottle
Catalog number 425902500
Price (USD)
144.65
Online Exclusive
161.00
Save 16.35 (10%)
Each
Quantity:
250 mg
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
144.65
Online Exclusive
161.00
Save 16.35 (10%)
Each
Chemical Identifiers
CAS22255-16-9
IUPAC Name(1R,2R,5S)-3-azabicyclo[3.1.0]hexan-3-ium-2-carboxylate
Molecular FormulaC6H9NO2
InChI KeyJBDOTWVUXVXVDR-UOWFLXDJSA-N
SMILES[O-]C(=O)[C@@H]1[NH2+]C[C@H]2C[C@@H]12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder or crystalline powder
Infrared spectrumConforms
Titration with NaOH>=94 %
NMR (1H-NMR)Consistent with structure
Appearance (Color)White to yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only