Bromine, 1M solution in trimethyl phosphate, AcroSeal™, Thermo Scientific Chemicals
Bromine, 1M solution in trimethyl phosphate, AcroSeal™, Thermo Scientific Chemicals
Bromine, 1M solution in trimethyl phosphate, AcroSeal™, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Bromine, 1M solution in trimethyl phosphate, AcroSeal™, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantityPackaging
428721000100 mLAcroSeal™ Glass Bottle
Catalog number 428721000
Price (USD)
-
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS7726-95-6
IUPAC Namebromide
Molecular FormulaBr
InChI KeyCPELXLSAUQHCOX-UHFFFAOYSA-M
SMILES[Br-]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Orange to red to brown
Titration with Na2S2O30.9 to 1.5 M
Appearance (Form)Solution
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only