TaqMan™ Exogenous Internal Positive Control Reagents, 200 reactions - FAQs
TaqMan™ Exogenous Internal Positive Control Reagents, 200 reactions - FAQs
View additional product information for TaqMan™ Exogenous Internal Positive Control Reagents - FAQs (4308321, 4308323)
1 product FAQs found
Can I order TaqMan Exogenous Internal Positive Control Reagents (Cat. Nos. 4308321, 4308323) with a different quencher?
Yes, you can order TaqMan Exogenous Internal Positive Control Reagents with QSY through Specialty Oligos with no need for redesign. You may reach out to our Specialty Oligos team at Specialty_Oligos@thermofisher.com.
You can also order them with MGB, but this would require redesign, and they would need to be made with bioinformatics through the Assay Design Hub, if possible.
Find additional tips, troubleshooting help, and resources within our TaqMan Primers and Probes Support Center.