Try PowerTrack SYBR Green Master Mix - our newest formulation simplifies your workflow by minimizing primer optimization and reducing pipetting errors with an optional sample tracking dye.
SYBR™ Green Universal Master Mix
Applied Biosystems™

SYBR™ Green Universal Master Mix

Notification of updated SYBR Green master mix formulation: PowerTrack SYBR Green Master Mix is our newest formulation designed to greatlyRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
43091551 x 5 mL
43444631 x 1 mL
43643442 x 5 mL
43643465 x 5 mL
431270410 x 5 mL
43349731 x 50 mL
Catalog number 4309155
Price (USD)
594.65
Online exclusive
646.00
Save 51.35
Each
Quantity:
1 x 5 mL
Price (USD)
594.65
Online exclusive
646.00
Save 51.35
Each

Notification of updated SYBR Green master mix formulation: PowerTrack SYBR Green Master Mix is our newest formulation designed to greatly simplify your work and deliver high sensitivity and specificity. It comes with an optional yellow dye that can be added to your DNA sample for pipetting QC. Yellow sample added to blue master mix yields green qPCR reaction. Visually confirm that your DNA sample was added to master mix. PowerTrack SYBR Green Master Mix is compatible with a wide range of primer melting temperatures (55°C to 65°C) and concentrations to minimize primer optimization work.

----------------------------------------------

SYBR Green PCR Master Mix
Everything you need for SYBR Green dye–based PCR amplification and detection in a convenient, single-tube format. Applied Biosystems SYBR Green PCR Master Mix combines SYBR Green I dye, AmpliTaq Gold DNA Polymerase, dNTPs with dUTP, Passive Reference 1, and optimized buffer in the convenience of a single vial.

  • Premixed components stored at 2–8°C significantly reduce assay setup time
  • SYBR Green I dye detects double-stranded DNA, so specific probes are not required
  • AmpliTaq Gold™ DNA polymerase minimizes nonspecific product formation to achieve superior performance
  • dUTP significantly reduces carryover contamination when used in conjunction with uracil-DNA glycosylase
  • Proprietary buffer enhancements ensure performance and reliability

Maximum flexibility and convenience
Applied Biosystems SYBR Green PCR Master Mix provides maximum flexibility at reduced cost because no target-specific TaqMan probes are required. SYBR Green I dye is a double-stranded DNA binding dye that detects any double-stranded DNA generated during PCR. The hot-start enzyme AmpliTaq Gold DNA Polymerase minimizes nonspecific product formation (including primer-dimers), yielding superior performance and sensitivity. Passive Internal Reference 1 is provided to normalize non-PCR–related fluorescence fluctuations. This minimizes well-to-well variability that can result from a variety of causes, such as pipetting error or sample evaporation. SYBR Green I dye is ideal for target identification (screening assays) or when a limited number of assays is needed.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)7000 System, 7300 System, 7500 System, 7700 System, 7900HT System,Applied Biosystems StepOnePlus™ Fast Real-Time PCR System, QuantStudio™, StepOne™, Standard Mode, StepOnePlus™, Standard Mode, ViiA™ 7 System
FormatTube
No. of Reactions200 Reactions
Passive Reference DyeROX (Pre-mixed)
PolymeraseAmpliTaq Gold DNA Polymerase
Product LineSYBR
Product TypeReal Time PCR SYBR Master Mix
Quantity1 x 5 mL
Sample TypeDNA (Genomic), cDNA
Shipping ConditionWet Ice
Sufficient For200 Reactions of 50 μL
Volume1 x 5 mL
Concentration2X
Detection MethodSYBR
For Use With (Application)Gene Expression
GC-Rich PCR PerformanceHigh
Label or DyeSYBR Green
PCR MethodqPCR
Reaction SpeedStandard
Unit SizeEach
Contents & Storage
The 1 x 5 mL vials contain a 2X mixture of SYBR Green 1 Dye, AmpliTaq Gold™ DNA Polymerase, dNTPs with dUTP, Passive Reference 1 (ROX), and optimized buffer components. Sufficient reagents provided for 200 reactions based on a 50 μl reaction volume.

Store at -20°C.

Guaranteed minimum shelf life is 60 days (exact expiry date printed on product and CofA).

Frequently asked questions (FAQs)

Can I use SYBR Green PCR Master Mix (Cat. No. 4309155, 4364344, 4364346, 4312704, 4334973) in Fast mode?

SYBR Green PCR Master Mix is not supported for use with Fast Mode thermal cycling conditions. When using SYBR Green PCR Master Mix on the StepOne, 7500 Fast, or 7900HT Fast instruments, use Standard mode thermal cycling conditions.

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Applications Support Center.

When using SYBR Green dye chemistry on an Applied Biosystems 7900HT Fast, a 7500 or 7500 Fast, or a 7300 Real-Time PCR System, is there an option to set the quencher to reflect the fact that there is no TaqMan probe being used in the reaction?

Yes. On the Detector Manager for the System, set the Quencher to “Non Fluorescent”.

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Applications Support Center.

Is the use of AmpErase UNG necessary for performing reverse transcription reactions?

No. We do not recommend that you use AmpErase UNG when performing reverse transcription. When using a dNTP mix with dUTP in a RT reaction, uracil will get incorporated into the cDNA generated from your RNA template. AmpErase UNG (uracil N-glycosylase), through an enzymatic reaction, will cleave single or double stranded DNA dUTP containing sequences.

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Applications Support Center.

Can Power SYBR Green PCR Master Mix (Cat. No. 4368577, 4367659, 4368706, 4368702, 4368708, 4367660) be substituted directly for the SYBR Green PCR Master Mix (Cat. No. 4309155, 4364344, 4364346, 4312704, 4334973)?

The Power SYBR Green PCR Master Mix is a new and improved formulation of the regular SYBR Green PCR Master Mix. You may need to verify your assays in order to start using the Power SYBR Green PCR Master Mix. For more information on verifying your reaction with the Power SYBR Green PCR Master Mix, please refer to the Power SYBR Green PCR Master Mix protocol.

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Applications Support Center.

Does the SYBR Green PCR Master Mix (P/N 4309155) contain AmpErase UNG?

SYBR Green PCR Master Mix (P/N 4309155) does not contain AmpErase UNG. It contains the following components: SYBR Green 1 dye, AmpliTaq Gold DNA polymerase, dNTPs (with dUTP), Passive Reference 1 and optimized buffer components; all the necessary components to perform real-time PCR in a convenient premixed 2X solution. This product has been tested on and is supported for use on an ABI PRISM Sequence Detection System. For more information, please consult the SYBR Green PCR Master Mix protocol.

Citations & References (1)

Citations & References
Abstract
Pharmacological ER stress promotes hepatic lipogenesis and lipid droplet formation.
Authors:Lee JS, Mendez R, Heng HH, Yang ZQ, Zhang K,
Journal:Am J Transl Res
PubMed ID:22347525
Endoplasmic Reticulum (ER) stress refers to a condition of accumulation of unfolded or misfolded proteins in the ER lumen. A variety of biochemical stimuli or pathophysiologic conditions can directly or indirectly induce ER stress, leading to activation of an ER-originated adaptive signaling response called Unfolded Protein Response (UPR). Recent studies ... More