SNaPshot™ Multiplex Kit, with protocol, 100 reactions - FAQs
SNaPshot™ Multiplex Kit, with protocol, 100 reactions - FAQs
View additional product information for SNaPshot™ Multiplex Kit - FAQs (4323159, 4323163, 4323151, 4323161)
1 product FAQs found
How many SNPs can the SNaPshot Multiplex kit analyze?
The SNaPshot Multiplex Kis is designed to interrogate up to 10 single nucleotide polymorphisms (SNPs) at known locations on 1-10 templates in a single tube.