TaqMan™ Universal PCR Master Mix, no AmpErase™ UNG
Applied Biosystems™

TaqMan™ Universal PCR Master Mix, no AmpErase™ UNG

TaqMan Universal PCR Master Mix, No AmpErase UNG, is the ideal reagent solution when you need a master mix forRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
432402010 x 5 mL, 2,000 reactions
43240181 x 5 mL, 200 reactions
43266141 x 50 mL 2,000 reactions
43643412 x 5 mL, 400 reactions
43643435 x 5 mL, 1,000 reactions
Catalog number 4324020
Price (USD)
6,115.65
Online exclusive
6,720.00
Save 604.35
Each
Quantity:
10 x 5 mL, 2,000 reactions
Price (USD)
6,115.65
Online exclusive
6,720.00
Save 604.35
Each
TaqMan Universal PCR Master Mix, No AmpErase UNG, is the ideal reagent solution when you need a master mix for multiple 5' nuclease DNA applications. Applied Biosystems reagents have been validated with TaqMan assays and Applied Biosystems real-time systems to ensure sensitive, accurate, and reliable performance every time.

• Contains AmpliTaq Gold DNA Polymerase to provide a better yield and a more robust 5' nuclease assay than AmpliTaq DNA Polymerase
• Includes a passive internal reference based on proprietary ROX dye, for superb precision on Applied Biosystems real-time PCR instruments
• Buffer enhancements guarantee performance and reliability in all applications, even with G/C-rich sequences
• Contains dUTP in the dNTP mix to protect against carry-over contamination, when used with AmpErase UNG
• Rapid assay development guidelines are provided to minimize optimization time

Alternative product: Try TaqMan Fast Advanced Master Mix, our highest-performance, probe-based master mix. With TaqMan Fast Advanced Master Mix, we’ve taken the best of TaqMan Universal PCR Master Mix and added additional capabilities for your gene expression analysis.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)7000 System, 7300 System, 7500 System, 7700 System, 7900HT System,Applied Biosystems StepOnePlus™ Fast Real-Time PCR System, GeneAmp 9600, StepOne™, Standard Mode, StepOnePlus™, Standard Mode
No. of Reactions2000 Reactions
Passive Reference DyeROX (Pre-mixed)
PolymeraseAmpliTaq Gold DNA Polymerase
Product LineTaqMan
Product TypeUniversal PCR Master Mix
Quantity10 x 5 mL, 2,000 reactions
Sample TypeDNA (Genomic), cDNA
Shipping ConditionWet Ice
Sufficient For2000 Reactions
Volume50 mL
Detection MethodPrimer-probe
For Use With (Application)Gene Expression, Genotyping, miRNA analysis
GC-Rich PCR PerformanceHigh
PCR MethodqPCR
Reaction SpeedStandard
Unit SizeEach
Contents & Storage
Supplied at 2X concentration. The mix contains AmpliTaq Gold™ DNA Polymerase, dNTPs with dUTP, Passive Reference 1 and optimized buffer components. This pack contains 10 X 5 mL tubes, sufficient for 2000 reactions at the 50 μL total volume. Store at 2-8°C

Guaranteed minimum shelf life is 60 days (exact expiry date printed on product and CofA).

Frequently asked questions (FAQs)

What master mix can I use with the TaqMan SNP Genotyping Assays?

We recommend using either the TaqPath ProAmp Master Mix or the TaqMan Genotyping Master Mix. The TaqMan Genotyping Master Mix has the advantage of proven performance with up to 3 days of pre- and post-PCR stability, allowing you to set up plates ahead of time or read the plates later (see the data here, https://tools.thermofisher.com/content/sfs/brochures/cms_039236.pdf) while the TaqPath ProAmp Master Mix can handle samples that may have inhibitors present.