Custom TaqMan™ Gene Expression Assay, FAM
Applied Biosystems™

Custom TaqMan™ Gene Expression Assay, FAM

Applied Biosystems Custom TaqMan Gene Expression assays are the custom version of our predesigned TaqMan assays for quantitative real-time PCR.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
4331348CNS (360 Reactions), Made to Order
4332079CNL (2900 Reactions), Made to Order
Catalog number 4331348CN
Price (USD)
-
Quantity:
S (360 Reactions), Made to Order

Applied Biosystems Custom TaqMan Gene Expression assays are the custom version of our predesigned TaqMan assays for quantitative real-time PCR. The Custom TaqMan Gene Expression assays consist of a pair of unlabeled PCR primers and a TaqMan probe with a dye label (FAM) on the 5' end and a minor groove binder (MGB) and nonfluorescent quencher (NFQ) on the 3' end.

Features include:

  • Easy to use—just add cDNA and master mix and run the qPCR—no melt curves required
  • Specific—TaqMan assays use proprietary MGB-containing probes that can be up to 15 bases shorter than non-MGB probes, improving the specificity of the assay
  • Sensitive—TaqMan assays are ideal for measuring low levels of expression or low-abundance targets
  • Accurate—identify small fold-changes with high accuracy of quantitation
  • Gold-standard TaqMan qPCR chemistry—TaqMan assays draw on Thermo Fisher Scientific's bioinformatics assay design pipeline to help ensure high specificity and minimal cross-reactivity, even for gene variants with high sequence homology
  • Sequences provided with every Custom TaqMan Gene Expression Assay you design and order

Approximate ship time

Made to order: 4–6 days in North America, 4–8 days in Europe

Pre-formulated assay

  • 2 unlabeled PCR primers (900 nM each final 1X concentration)
  • 1 FAM dye-labeled TaqMan MGB probe (250 nM final 1X concentration)

TaqMan Gene Expression assays are the gold standard in real-time PCR gene expression studies, built on more than 20 years of experience. Each assay includes target primers and a sequence-specific probe optimized for the best functional performance. No additional design, optimization, or melt curve analysis is needed. Available in a wide variety of formats and species, new assay designs are constantly added to help meet your research needs. TaqMan assays have been cited in over 40,000 publications and are backed by more than 350 patents.

Recommended master mix: TaqMan Fast Advanced Master Mix (Cat. No. 4444556)

See also: Custom Plus TaqMan RNA Assays, designed using the full bioinformatic power of the TaqMan Assay design pipeline.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)7500 System, 7500 Fast System, 7900HT System, StepOne™, StepOnePlus™, ViiA™ 7 System, QuantStudio™ 3, QuantStudio™ 5, QuantStudio™ 6 Flex, QuantStudio™ 7 Flex, QuantStudio 6 Pro, QuantStudio 7 Pro, QuantStudio™ 12k Flex
No. of Reactions360 Reactions, Made to Order
Product LineTaqMan
Product TypeCustom TaqMan Gene Expression
QuantityS (360 Reactions), Made to Order
Shipping ConditionRoom Temperature
For Use With (Application)Gene Expression
Unit SizeEach
Contents & Storage
1 tube containing a 20X (S and M sizes) or 60X (L size) mix of pre-formulated assay (1 probe and 2 primers).

Store at -15 to -25°C.