MicroSEQ™ D2 rDNA Fungal Sequencing Kit - FAQs
MicroSEQ™ D2 rDNA Fungal Sequencing Kit - FAQs
View additional product information for MicroSEQ™ D2 rDNA Fungal Sequencing Kit - FAQs (4347481)
1 product FAQs found
Do you have stability information for the MicroSEQ D2 rDNA Fungal Sequencing Kit (Cat. No. 4347481) when subjected to temperature fluctuations?
The MicroSEQ D2 rDNA Fungal Sequencing Kit is stable if stored at -20 degrees C for 15-16 months. We do not have any information available for temperatures outside of this range.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Applications Support Center.