MicroSEQ™ 500 16S rDNA PCR Kit - FAQs
MicroSEQ™ 500 16S rDNA PCR Kit - FAQs
View additional product information for MicroSEQ™ 500 16S rDNA PCR Kit - FAQs (4348228)
1 product FAQs found
What is the sensitivity of the MicroSeq 500 kits?
As long as you rstart from a visible colony or cell pellet, MicroSeq kits will work.