7500 Fast Real-Time PCR Systems Spectral Calibration Kit I - FAQs
7500 Fast Real-Time PCR Systems Spectral Calibration Kit I - FAQs
View additional product information for 7500 Fast Real-Time PCR Systems Spectral Calibration Kit I - FAQs (4360788)
1 product FAQs found
Can I use the 7500 Fast Real-Time PCR Systems Spectral Calibration Kit I (Cat. No. 4360788) with the QuantStudio 5 Real-Time PCR System?
The 7500 Fast Real-Time PCR Systems Spectral Calibration Kit I (Cat. No. 4360788) has been validated to be used only with the 7500 Fast Real-Time PCR instrument with a 0.1 mL block. The part numbers for the spectral calibration kits to be used with QuantStudio 5 Real-Time PCR System (96 well, 0.2 mL or 0.1 mL) are as follows:
  • QuantStudio 3/5 Spectral Calibration Plate 1 (Cat. No. A26331)
  • QuantStudio 3/5 Spectral Calibration Plate 2, 96 Well 0.2 mL (Cat. No. A26332)
  • 96 Well 0.1 mL Spectral Calibration Plate 1 (Cat. No. A26336)
  • 96 Well 0.1 mL Spectral Calibration Plate 2 (Cat. No. A26337)
  • 96 Well 0.1 mL Spectral Calibration Plate 3 (Cat. No. A26340)
  • QuantStudio 3 or 5 10‑Dye Spectral Calibration Kit, 96 Well 0.1 mL (Cat. No. A26342)
  • QuantStudio 3/5 Spectral Calibration Plate 3, 96 Well 0.2 mL (Cat. No. A26333)
  • QuantStudio 3/5 10‑Dye Spectral Calibration Kit, 96 Well 0.2 mL (Cat. No. A26343)
Find additional tips, troubleshooting help, and resources within our QuantStudio 3 and QuantStudio 5 Real Time PCR Systems Support Center