Acetobromo-alpha-D-glucuronic acid methyl ester, 98%
Acetobromo-alpha-D-glucuronic acid methyl ester, 98%
Acetobromo-alpha-D-glucuronic acid methyl ester, 98%
Thermo Scientific Chemicals

Acetobromo-alpha-D-glucuronic acid methyl ester, 98%

CAS: 21085-72-3 | C13H17BrO9 | 397.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 436290010
Price (USD)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS21085-72-3
IUPAC Namemethyl (2S,3S,4S,5R,6R)-3,4,5-tris(acetyloxy)-6-bromooxane-2-carboxylate
Molecular FormulaC13H17BrO9
InChI KeyGWTNLHGTLIBHHZ-SVNGYHJRSA-N
SMILESCOC(=O)[C@H]1O[C@H](Br)[C@H](OC(C)=O)[C@@H](OC(C)=O)[C@@H]1OC(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Stabilizer2 % CaCO3
Appearance (Form)Crystalline powder or solid
HPLC>=97.5 %
Appearance (Color)White to grey
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only