MicroAmp™ Fast Optical 96-Well Reaction Plate with Barcode, 0.1 mL, 200 Plates - FAQs
MicroAmp™ Fast Optical 96-Well Reaction Plate with Barcode, 0.1 mL, 200 Plates - FAQs
View additional product information for MicroAmp™ Fast Optical 96-Well Reaction Plate with Barcode, 0.1 mL - FAQs (4366932, 4346906)
3 product FAQs found
Are the MicroAmp Optical 96-Well Reaction Plates and MicroAmp Optical 384 -Well Reaction Plates autoclavable?
Our microplates are certified to be DNA, RNase and PCR inhibitor free as long as they are kept in the original packaging bag. They are not meant to be autoclaved.
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.
Can I place an Optical Fast 96-Well plate onto a standard Applied Biosystems 7500 or 7300 Real-Time PCR System?
No. The fast plate will not fit correctly into the standard block and assay performance will be significantly compromised.
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.
Can I use MicroAmp Fast Optical 96-Well Reaction Plate with Barcode, 0.1 mL (Cat. No. 4346906, 4366932) with the QuantStudio 7 Flex Real-Time PCR System in the Standard run mode?
Yes. MicroAmp Fast Optical 96-Well Reaction Plate with Barcode, 0.1 mL (Cat. No. 4346906, 4366932) can be used with either Fast or Standard run modes. However, the “Fast” plates can only be run on the 96-well Fast block, which is for smaller volumes rather than a standard block (0.2 mL). The Fast or Standard mode for the run is determined by the polymerase in the master mix used in the experiment. Please refer to the user guide for the specific master mix to determine which mode to use.
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.