Not what you're looking for? Search all of our TaqMan® arrays for your gene or pathway of interest.
Do you need this panel in a 384-well plate? Please visit Specialty Plating to learn more.
Product Image
Applied Biosystems™

TaqMan™ Array Human Protein Kinase Panel

The Applied Biosystems™ TaqMan™ Human Protein Kinase Array is an efficient, easy-to-use micro fluidic card for quantitative gene expression analysis of human protein kinase genes important in drug discovery.
Have Questions?
Catalog NumberQuantity
43677844 cards
Catalog number 4367784
Price (USD)
2,635.00
Each
Quantity:
4 cards
Price (USD)
2,635.00
Each

The Applied Biosystems™ TaqMan™ Human Protein Kinase Array is an efficient, easy-to-use micro fluidic card for quantitative gene expression analysis of human protein kinase genes important in drug discovery. This product is designed with TaqMan™ Gene Expression Assays arrayed in singletons in 384 wells and is compatible with an upgraded Applied Biosystems 7900HT Fast Real-Time PCR System.

The Human Protein Kinase Array contains assays for 68 human kinases. Fifty-three of the 68 kinases are from receptor protein-tyrosine kinase (RPTK) families: EGFR, InsulinR, PDGFR, VEGFR, FGFR, CCK, NGFR, HGFR, EPHR, AXL, TIE, RYK, DDR, RET, ROS, LTK, ROR and MUSK. The remaining 15 kinases are Ser/Thr kinases from the kinase families: CAMKL, IRAK, Lmr, RIPK and STKR. We have also selected assays for 26 classic endogenous, non-kinase genes. These genes are involved in signal transduction and mediate protein-protein interaction, transcriptional regulation, neural development and cell adhesion. This product is sold in packages of 4 cards.

The array is pre-formatted and inventoried, cost-effective, convenient and easy to use without needing expensive robotics. Results are reproducible and consistent across samples, studies, and labs —the same data quality from card-to-card and lot-to-lot — even with different operators.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Format384-well microfluidic card
Internal Probe ModificationMGB (Minor Groove Binder) (3'), Non-Fluorescent Quencher (3'), FAM (5')
No. of Reactions1 x 2 μL Reactions
Product LineTaqMan
Quantity4 cards
Research CategoryCell Cycle & Proliferation, Apoptosis, Neurobiology, Signal Transduction, Oncology, Biomarker-Related Pathway(s), Extracellular Matrix & Cell Adhesion, Reproduction, Toxicology & Drug Metabolism, Immunology, Development
SpeciesHuman
Target68 human kinases
For Use With (Application)Gene Expression
FormDried Down
Label or DyeFAM
Reaction SpeedStandard
Unit SizeEach
Contents & Storage
Each TaqMan™ Array well contains a single TaqMan™ Assay for one 2 μL reaction. The cards should be stored at 4°C.

Frequently asked questions (FAQs)

What are the storage recommendations for TaqMan Array Cards (TAC)?

We recommend storing TaqMan Array Cards (TAC) at 2-8 degrees C. TaqMan Array Cards (TAC) are stable for 2 years from the manufacturing date. The expiration date is listed on the packaging.

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.