Do you have stability information for the FAST MicroSEQ 500 16S rDNA PCR Kit (Cat. No. 4370489) when subjected to temperature fluctuations?
The FAST MicroSEQ 500 16S rDNA PCR Kit is stable if stored at -20 degrees C for 15-16 months. We do not have any information available for temperatures outside of this range.