MicroSEQ™ ID 16S rDNA Full Gene Library v2.0 - FAQs
MicroSEQ™ ID 16S rDNA Full Gene Library v2.0 - FAQs
View additional product information for MicroSEQ™ ID 16S rDNA Full Gene Library v2.0 - FAQs (4371295)
2 product FAQs found
You offer MicroSEQ ID 16S rDNA Full Gene Library v2.0, USB format (Cat. No. A46365) and MicroSEQ ID 16S rDNA Full Gene Library v2.0 (Cat. No. 4371295). What is the difference between them?
The only difference is that Cat. No. A46365 is available in USB format, otherwise the content is the same between the two catalog numbers.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.
What is the difference in library format between MicroSEQ ID 16S rDNA Full Gene Library v2.0 and MicroSEQ ID 16S rDNA Full Gene Library v1.0?
MicroSEQ ID 16S rDNA Full Gene Library v2.0 is the same as MicroSEQ ID 16S rDNA Full Gene Library 1.0 but in USB format. No new content has been added.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.