High-Capacity cDNA Reverse Transcription Kit with RNase Inhibitor, 200 reactions - FAQs
High-Capacity cDNA Reverse Transcription Kit with RNase Inhibitor, 200 reactions - FAQs
View additional product information for High-Capacity cDNA Reverse Transcription Kit - FAQs (4368814, 4374967, 4374966, 4368813)
3 product FAQs found
Can I order the RNase Inhibitor from the High-Capacity cDNA Reverse Transcription Kit separately?
Yes. You can order the RNase Inhibitor separately as Cat. No. N8080119.
Can one use the reagents from High Capacity RT kit to make cDNA for TaqMan MicroRNA Assay?
Yes. You can use the reagents from the High Capacity cDNA Kit PN 4368814 (without RNAse Inhibitor) or 4374966 (with RNAse Inhibitor). Make sure to use the reverse transcription primer provided from the TaqMan miRNA assay for the reverse transcription.
Which kit should I use for the reverse transcription step for TaqMan Pri-miRNA assays (Cat. No. 4427012)?
We recommend the following kits:
  • High-Capacity cDNA Reverse Transcription Kit (Cat. No. 4374966).
  • High-Capacity RNA-to-cDNA Kit (Cat. No. 4387406).
Find additional tips, troubleshooting help, and resources within our PCR and cDNA Synthesis Support Center.