Can I order the RNase Inhibitor from the High-Capacity cDNA Reverse Transcription Kit separately?
Yes. You can order the RNase Inhibitor separately as Cat. No. N8080119.
Search
View additional product information for High-Capacity cDNA Reverse Transcription Kit - FAQs (4368814, 4374967, 4374966, 4368813)
2 product FAQs found