MicroAmp™ Fast Optical 48-Well Reaction Plate - FAQs
MicroAmp™ Fast Optical 48-Well Reaction Plate - FAQs
View additional product information for MicroAmp™ Fast Optical 48-Well Reaction Plate - FAQs (4375816)
1 product FAQs found
Can I cut a 96-well FAST plate to fit the StepOne system?
No, the MicroAmp Fast Optical 48-Well Reaction Plate (P/N 4375816) is the only plate that can be used on the StepOne system. Using a plate that has not been designed for use with the StepOne Real-Time PCR System is not recommended and can potentially damage the instrument.
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.