BOC-D-alpha-phenylglycine, 99%
BOC-D-alpha-phenylglycine, 99%
BOC-D-alpha-phenylglycine, 99%
Thermo Scientific Chemicals

BOC-D-alpha-phenylglycine, 99%

CAS: 33125-05-2 | C13H17NO4 | 251.28 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 437640050
Price (USD)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS33125-05-2
IUPAC Name2-{[(tert-butoxy)carbonyl]amino}-2-phenylacetic acid
Molecular FormulaC13H17NO4
InChI KeyHOBFSNNENNQQIU-UHFFFAOYNA-N
SMILESCC(C)(C)OC(=O)NC(C(O)=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Specific optical rotation-142° to -146° (20°C, 589 nm) (c=1, ethanol)
Appearance (Color)White
Appearance (Form)Powder or crystalline powder
Infrared spectrumConforms
HPLC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only