Not what you're looking for? Search all of our TaqMan® arrays for your gene or pathway of interest.
Do you need this panel in a 384-well plate? Please visit Specialty Plating to learn more.
TaqMan™ Array Human Apoptosis Panel
Applied Biosystems™

TaqMan™ Array Human Apoptosis Panel

The Applied Biosystems™ TaqMan™ Human Apoptosis Array is an efficient, easy-to-use micro fluidic card for quantitative gene expression analysis of targets known to have implications in programmed cell death.
Have Questions?
Catalog NumberQuantity
43787014 cards
Catalog number 4378701
Price (USD)
2,590.00
Each
Quantity:
4 cards
Price (USD)
2,590.00
Each

The Applied Biosystems™ TaqMan™ Human Apoptosis Array is an efficient, easy-to-use micro fluidic card for quantitative gene expression analysis of targets known to have implications in programmed cell death. The study of apoptosis is important in cancer, HIV, and other disease research. This product is designed with 96 TaqMan™ Gene Expression Assays (93 targets & 3 controls) loaded in singletons into the array’s 384 wells and is compatible with an upgraded Applied Biosystems 7900HT Fast Real-Time PCR System. This product is packaged in 1 box containing 4 cards.

The TaqMan™ Human Apoptosis Array contains assays for 93 human genes in addition to 3 endogenous controls (18S, ACTB, GAPDH). The 93 genes are categorized in multiple target classes or pathways. The array is pre-formatted and inventoried, cost-effective, convenient and easy to use without needing expensive robotics. Results are reproducible and consistent across samples, studies, and labs —the same data quality from card-to-card and lot-to-lot — even with different operators.

For Research Use Only. Not for use in diagnostic procedures.

Specifications
Format384-well microfluidic card
Internal Probe ModificationMGB (Minor Groove Binder) (3'), Non-Fluorescent Quencher (3'), FAM (5')
No. of Reactions1 x 2 μL Reactions
Product LineTaqMan
Quantity4 cards
Research CategoryCell Cycle & Proliferation, Apoptosis, Neurobiology, Signal Transduction, Biomarker-Related Pathway(s), Extracellular Matrix & Cell Adhesion, Reproduction, Toxicology & Drug Metabolism, Immunology, Oncology
SpeciesHuman
Target93 human apoptosis-related genes
For Use With (Application)Oncology, Genetic Disease Research
FormDried Down
Label or DyeFAM
Reaction SpeedStandard
Unit SizeEach
Contents & Storage
Each TaqMan™ Array well contains a single TaqMan™ Assay for one 2 μL reaction. The cards should be stored at 4°C.

Frequently asked questions (FAQs)

What are the storage recommendations for TaqMan Array Cards (TAC)?

We recommend storing TaqMan Array Cards (TAC) at 2-8 degrees C. TaqMan Array Cards (TAC) are stable for 2 years from the manufacturing date. The expiration date is listed on the packaging.

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.