Not what you're looking for? Search all of our TaqMan® arrays for your gene or pathway of interest.
Do you need this panel in a 384-well plate? Please visit Specialty Plating to learn more.
TaqMan™ Array Rat Endogenous Control Panel
Applied Biosystems™

TaqMan™ Array Rat Endogenous Control Panel

The Applied Biosystems™ TaqMan™ Rat Endogenous Control Array is a 384-well micro flluidic card containing 16 rat TaqMan™ Gene Expression Assays commonly used as endogenous controls arrayed in triplicates.
Have Questions?
Catalog NumberQuantity
43787042 cards
Catalog number 4378704
Price (USD)
1,194.00
Each
Quantity:
2 cards
Price (USD)
1,194.00
Each

The Applied Biosystems™ TaqMan™ Rat Endogenous Control Array is a 384-well micro flluidic card containing 16 rat TaqMan™ Gene Expression Assays commonly used as endogenous controls arrayed in triplicates. This product is compatible with an upgraded Applied Biosystems 7900HT Fast Real-Time PCR System. Endogenous Control Panels are sold in packs of 2.

The array is pre-formatted and inventoried, cost-effective, convenient and easy to use without needing expensive robotics. Results are reproducible and consistent across samples, studies, and labs —the same data quality from card-to-card and lot-to-lot — even with different operators.

For Research Use Only. Not for use in diagnostic procedures.

Specifications
Description16 assays for housekeeping genes and genes with minimal differential expression
Format384-well microfluidic card
Internal Probe ModificationMGB (Minor Groove Binder) (3'), Non-Fluorescent Quencher (3'), FAM (5')
Product LineTaqMan
Quantity2 cards
SpeciesRat
TargetHousekeeping & Control Gene Panel
For Use With (Application)Gene Expression
FormDried Down
Label or DyeFAM
Reaction SpeedStandard
Unit SizeEach
Contents & Storage
Control Primer-Probe Set(s)

Frequently asked questions (FAQs)

What are the storage recommendations for TaqMan Array Cards (TAC)?

We recommend storing TaqMan Array Cards (TAC) at 2-8 degrees C. TaqMan Array Cards (TAC) are stable for 2 years from the manufacturing date. The expiration date is listed on the packaging.

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.

Can poly A+ purified RNA samples be used with the TaqMan Human Endogenous Control Plate (Cta. No. 4309199)?

No. The plate has been designed to evaluate only total RNA. Ribosomal RNA will be removed from poly A+ purified RNA samples, and is required for accurate detection of the 18s rRNA endogenous control assay used on the plate.