Tempus™ Spin RNA Isolation Kit - FAQs
Tempus™ Spin RNA Isolation Kit - FAQs
View additional product information for Tempus™ Spin RNA Isolation Kit - FAQs (4380204)
4 product FAQs found
Will the Tempus system work to isolate miRNA?
No, unfortunately, small RNAs will be lost using this purification method due to the low molecular weight cutoff from the column (approximately 200 bp).
I am doing blood work and using the Tempus collection tubes. Is the Tempus system compatible with the RiboPure-Blood RNA Purification Kit?
No, please follow the RNA isolation methods described with the Tempus system. The collection tubes are not compatible with the RiboPure kit.
What is the recommended storage temperature and stability of samples in the Tempus Blood RNA Tube?
Our general recommendations are that once blood has been collected in the Tempus Blood RNA Tubes, you can store the tube at room temperature for up to 5 days or at 4 degrees C for up to 7 days. For long term storage, the tube can be frozen at -80 degrees C for at least 1.5 years.
Can I add a DNase step to the RNA isolation protocol in the Tempus Spin RNA Isolation Kit?
Yes, it is okay to add an on-column or post-elution DNase treatment.
For the on-column DNase treatment, use 350 µl of the following solution and incubate for 15 mins at room temperature:
2500 units DNAse I (RNAse-free) in 10 mL of solution containing:
10 mM Tris-HCl, pH 8
2.5 mM MgCl2 . 6H2O
0.1 mM CaCl2 . 2H2O
This would be done between the two Wash 2 steps, and it would be a good idea to add a third Wash 2 step to make sure all of the DNase is washed away.