I ran out of carrier RNA for my MagMAX isolation kit. Can I order this part separately?
Yes, carrier RNA can be ordered separately. The Cat. No. is 4382878.
Search
View additional product information for Carrier RNA - FAQs (4382878)
1 product FAQs found