Can I use preamplification with the TaqMan SNP Genotyping Assays?
There is a preamplification protocol included with our TaqMan Sample-to-SNP Kit. This is designed to work with a lysate sample.
Search
View additional product information for TaqMan™ PreAmp Master Mix Kit - FAQs (4384267)
4 product FAQs found
For more detailed information on this kit, please refer to the TaqMan PreAmp Master Mix Kit Protocol (P/N 4384557).