BOC-L-alaninal, 98%
BOC-L-alaninal, 98%
BOC-L-alaninal, 98%
Thermo Scientific Chemicals

BOC-L-alaninal, 98%

CAS: 79069-50-4 | C8H15NO3 | 173.21 g/mol
Have Questions?
Catalog NumberQuantity
4384700101 g
Catalog number 438470010
Price (USD)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS79069-50-4
IUPAC Nametert-butyl N-[(2S)-1-oxopropan-2-yl]carbamate
Molecular FormulaC8H15NO3
InChI KeyOEQRZPWMXXJEKU-LURJTMIESA-N
SMILESC[C@H](NC(=O)OC(C)(C)C)C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Powder
Specific optical rotation-29° to -39° (20°C, 589 nm) (C=2 in methanol)
Appearance (Color)White to yellow
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only