Product Image
Applied Biosystems™

TaqMan™ Array Mouse Stem Cell Pluripotency Panel

The Applied Biosystems™ TaqMan™ Mouse Stem Cell Pluripotency Array is an efficient, easy-to-use micro fluidic card for the characterization ofRead more
Catalog NumberQuantity
43853634 cards
Catalog number 4385363
Price (GBP)
1,352.00
Each
Quantity:
4 cards
Price (GBP)
1,352.00
Each

The Applied Biosystems™ TaqMan™ Mouse Stem Cell Pluripotency Array is an efficient, easy-to-use micro fluidic card for the characterization of undifferentiated stem cells or their differentiated derivatives. This gene signature panel contains mouse orthologs of well-defined human markers for pluripotency plus 6 endogenous controls. The format of the TaqMan Array allows for 1-4 replicates to be run in parallel per array. Each order includes a CD with assay information and an instrument software input file. This product is sold in packages of 4 cards.

The Mouse Stem Cell Pluripotency Array contains mouse ortholog genes of the human panel to characterize mouse embryonic stem cells. The gene content on this TaqMan Array resulted directly from the work of the I.S.C.I consortium and characterization of human embryonic stem cell lines by the International Stem Cell Initiative.

The array is pre-formatted and inventoried, cost-effective, convenient and easy to use without needing expensive robotics. Results are reproducible and consistent across samples, studies, and labs – the same data quality from card-to-card and lot-to-lot – even with different operators.

The Right Targets

These TaqMan Arrays use Applied Biosystem's proprietary micro fluidics technology to simultaneously analyze gene expression levels by quantitative PCR for 92 test genes and 6 endogenous controls. Genes were included based on the following criteria:

  1. Expression in human embryonal carcinoma cells (hEC), primordial germ cells or human ES (hES) cells
  2. Known role in maintenance of pluripotency in hES cells
  3. Identification in microarray-based experiments as being correlated with the stem cell state
  4. Expression known to be characteristic of specific differentiation lineages in the post implantation embryo and mature tissues. These are the right targets to characterize embryonic stem cells.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Format384-well microfluidic card
Internal Probe ModificationMGB (Minor Groove Binder) (3'), Non-Fluorescent Quencher (3'), FAM (5')
No. of Reactions1 x 2 μL Reactions
Product LineTaqMan
Quantity4 cards
Research CategoryCell Cycle & Proliferation, Biomarker-Related Pathway(s), Stem Cell Research, Reproduction
SpeciesMouse
TargetMouse orthologs of well-defined human markers for stem cell pluripotency
For Use With (Application)Stem Cell Analysis
FormDried Down
Label or DyeFAM
Reaction SpeedStandard
Unit SizeEach
Contents & Storage
Each TaqMan™ Array well contains a single TaqMan™ Assay for one 2 μL reaction. The cards should be stored at 4°C.

Frequently asked questions (FAQs)

What are the storage recommendations for TaqMan Array Cards (TAC)?

We recommend storing TaqMan Array Cards (TAC) at 2-8 degrees C. TaqMan Array Cards (TAC) are stable for 2 years from the manufacturing date. The expiration date is listed on the packaging.

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.