Fast SYBR™ Green Master Mix, 10 x 5 mL - FAQs
Fast SYBR™ Green Master Mix, 10 x 5 mL - FAQs
View additional product information for Fast SYBR™ Green Master Mix - FAQs (4385610, 4385612, 4385614, 4385616, 4385618, 4385617)
1 product FAQs found
Is there a SYBR Green Fast Master Mix currently available?
Yes. The following reagents can be used for fast cycling: Fast SYBR Green Master Mix, EXPRESS SYBR GreenER qPCR Supermix Universal, and SYBR GreenER qPCR SuperMix for ABI PRISM instruments.