Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Complex I Antibodies

          Invitrogen

          Complex I Monoclonal Antibody (18G12BC2)

          3 References

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite Complex I Monoclonal Antibody (18G12BC2)

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (3)
          Complex I Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          Complex I Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 3

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Complex I Antibody (43-8800) in ICC/IF

          Immunocytochemistry analysis of MT-ND1 in fibroblasts cells. The cells were fixed with 4% paraformaldehyde for 20 minutes, permeabilized with 0.1% Triton X-100 for 15 minutes, and incubated with MT-ND1 Monoclonal Antibody (Product # 43-8800) at 1 µg/mL for 2 hours at room temperature. 10% Goat serum was used as the blocking agent for all blocking steps. Detection was perfomed by an Alexa Fluor® 488 goat ant... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Complex I Antibody in Immunocytochemistry (ICC/IF)
          Complex I Antibody in Flow Cytometry (Flow)
          Complex I Antibody in Immunoprecipitation (IP)

          Product Details

          43-8800

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          -
          View 1 publication 1 publication

          Immunocytochemistry (ICC/IF)

          Assay-dependent
          -

          Flow Cytometry (Flow)

          Assay-dependent
          View 1 publication 1 publication

          Immunoprecipitation (IP)

          0.1-1.0 mg/mL
          -

          Proximity Ligation Assay (PLA) (PLA)

          -
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Bovine, Human, Mouse

          Published species

          Human, Mouse

          Host/Isotype

          Mouse / IgG2b, kappa

          Class

          Monoclonal

          Type

          Antibody

          Clone

          18G12BC2

          Immunogen

          Bovine Heart Complex I

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          IgG fraction

          Storage buffer

          HEPES buffered saline

          Contains

          0.02% sodium azide

          Storage conditions

          4°C, do not freeze

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2533543

          Product Specific Information

          Positive control: fibroblasts, HL-60 cells, tissue mitochondria.

          Target Information

          Complex I, or NADH ubiquinone oxidoreductase, is a large protein complex of 950,000 Da molecular weight made up by 45 to 46 different subunits. A total of seven of the subunits of the complex are encoded by mitochondrial DNA, while the remainder subunits are nuclear encoded, which are translated in the cytosol and translocated into the organelle for assembly at the inner membrane. The enzyme complex catalyses electron entry from NADH via a flavin (FMN) and several non-heme iron centers. Complex I is sensitive to a wide range of inhibitors, many of which are pesticides or other common environmental toxins, such as rotenone. Complex I dysfunction is a common cause of genetic OXPHOS defects. Altered functioning of this complex is also thought to contribute to several neurological disorders including Parkinson's disease and schizophrenia. Also, there is evidence of Complex I involvement in diabetes.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: 13 kDa differentiation-associated protein; 13kDa differentiation-associated protein; 14.5b subunit of the NADH-ubiquinone dehydrogenase complex; 16 kDa precursor; 19 kDa; 51 kda flavoprotein; ACP; Acyl carrier protein, mitochondrial; associated with retinoid-interferon induced mortality; B17.2-like; B17.2L; CD14; cell adhesion protein (SQM1); Cell adhesion protein SQM1; Cell death regulatory protein GRIM-19; cell death-regulatory protein GRIM19; CI-13kD-A; CI-13kD-B; CI-15 kDa; CI-18 kDa; CI-19kD; CI-20kD; CI-23kD; CI-30kD; CI-39kD; CI-42kD; CI-49kD; CI-51kD; CI-75kD; CI-9kD; CI-AGGG; CI-AQDQ; CI-ASHI; CI-B12; CI-B14; CI-B14.5a; CI-B14.5b; CI-B14.7; CI-B15; CI-B16.6; CI-B17; CI-B17.2; CI-B18; CI-B18; complex I-B18; CI-B22; CI-B8; CI-B9; CI-ESSS; CI-KFYI; CI-MLRQ; CI-MNLL; CI-MWFE; CI-PDSW; CI-PGIV; CI-SDAP; CI-SGDH; CIB17.2; complex 1, mitochondrial respiratory chain, 49-KD subunit; complex I 10kDa subunit; complex I 13kDa subunit A; complex I 13kDa subunit B; complex I 18kDa subunit; complex I 20kDa subunit; complex I 23kDa subunit; complex I 24kDa subunit; complex I 30kDa subunit; complex I 39kDa subunit; complex I 42kDa subunit; complex I 49kDa subunit; complex I 51 kda subunit; complex I 51kDa subunit; complex I 75kDa subunit; Complex I 9kDa subunit; complex I AGGG subunit; complex I ASHI subunit; complex I B14.5a subunit; complex I B14.7 subunit; complex I B15 subunit; complex I B16.6 subunit; complex I B17.2 subunit; complex I B18 subunit; complex I B22 subunit; complex I B8 subunit; complex I B9 subunit; Complex I intermediate-associated protein 30, mitochondrial; complex I iron sulphur protein subunit; 13kDa subunit; complex I iron sulphur protein subunit; 30 kDa subunit; complex I iron sulphur protein subunit; 49kDa subunit; complex I KFYI subunit; complex I MNLL subunit; complex I MWFE subunit; complex I NP17.3 subunit; complex I PDSW subunit; complex I SDAP subunit; complex I SGDH subunit; complex I subunit; Complex I subunit B13; complex I subunit B14.5b; complex I, mitochondrial respiratory chain; complex I, mitochondrial respiratory chain, 10-kD subunit; complex I, mitochondrial respiratory chain, 13-kD subunit; complex I, mitochondrial respiratory chain, 20-KD subunit; complex I, mitochondrial respiratory chain, 75-kD subunit; complex I, mitochondrial respiratory chain, B17 subunit; complex I, mitochondrial respitory chain, 24 kD subunit; Complex I-13kD-A; Complex I-13kD-B; Complex I-15 kDa; Complex I-18 kDa; Complex I-19kD; Complex I-20kD; Complex I-23kD; Complex I-30kD; Complex I-39kD; Complex I-42kD; Complex I-49kD; Complex I-51kD; Complex I-75kD; Complex I-9kD; Complex I-AGGG; Complex I-AQDQ; Complex I-ASHI; Complex I-B12; Complex I-B14; Complex I-B14.5a; Complex I-B14.5b; Complex I-B14.7; Complex I-B15; Complex I-B16.6; Complex I-B17; Complex I-B17.2; Complex I-B18; Complex I-B22; Complex I-B8; Complex I-B9; Complex I-ESSS; Complex I-KFYI; Complex I-MLRQ; Complex I-MNLL; Complex I-MWFE; Complex I-PDSW; Complex I-PGIV; Complex I-SGDH; component of mitochondrial Complex I; Cytochrome c oxidase hypoxia associated subunit FA4L2; Cytochrome c oxidase subunit FA4; cytochrome c oxidase subunit FA4 like 2; Cytochrome c oxidase subunit NDUFA4; DAP13; gene associated with retinoic and IFN-induced mortality 19 protein; Gene associated with retinoic and interferon-induced mortality 19 protein; Gene associated with retinoic-interferon-induced mortality 19 protein; genes associated with retinoid-IFN-induced mortality 19; GRIM-19; HLC-1; Hormone-regulated proliferation-associated protein of 20 kDa; Hormone-regulated proliferation-associated protein of 20 kDa homolog; hormone-regulated proliferation-associated protein, 20 kDa; Human lung cancer oncogene 1 protein; LOC56901; lung cancer oncogene 2; LYR motif-containing protein 3; LYR motif-containing protein 6; Mimitin; mimitin, mitochondrial; mitochondrial acyl carrier protein; mitochondrial NADH dehydrogenase ubiquinone flavoprotein 1; mitochondrial NADH oxidoreductase-like protein; mitochondrial NADH-ubiquinone oxidoreductase 75 kDa subunit; mitochondrial NADH:ubiquinone oxidoreductase 51 kda subunit; mitochondrial NADH:ubiquinone oxidoreductase B14.7; mitochondrial NADH:ubiquinone oxidoreductase ESSS subunit; mitochondrial respiratory chain associated factor 1; mitochondrial respiratory chain complex I (18-KD subunit); mitochondrial stress response 1; mitochondrial stress response hypoxia; MMTN; Myc-induced mitochondria protein; Myc-induced mitochondrial protein; NADH dehydrogenase (ubiquinone) 1 alpha subcomplex assembly factor 2 isoform NDUFAF2_1; NADH dehydrogenase (ubiquinone) 1 alpha subcomplex assembly factor 2 isoform NDUFAF2_3; NADH dehydrogenase (ubiquinone) 1 alpha subcomplex assembly factor 2 isoform NDUFAF2_4; NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 1 (7.5kD, MWFE); NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 1, 7.5kDa; NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 10, 42kDa; NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 11, 14.7kDa; NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 12; NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 13; NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 2, 8kDa; NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 3, 9kDa; NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 4 (9 kDa, MLRQ); NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 4, 9kDa; NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 4-like 2; NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 5, 13kDa; NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 6, 14kDa; NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 7, 14.5kDa; NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 8, 19kDa; NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 9, 39kDa; NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, assembly factor 1; NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, assembly factor 2; NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, assembly factor 3; NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, assembly factor 4; NADH dehydrogenase (ubiquinone) 1 assembly factor 1; NADH dehydrogenase (ubiquinone) 1 beta subcomplex, 1, 7kDa; NADH dehydrogenase (ubiquinone) 1 beta subcomplex, 10, 22kDa; NADH dehydrogenase (ubiquinone) 1 beta subcomplex, 11, 17.3kDa; NADH dehydrogenase (ubiquinone) 1 beta subcomplex, 2, 8kDa; NADH dehydrogenase (ubiquinone) 1 beta subcomplex, 3, 12kDa; NADH dehydrogenase (ubiquinone) 1 beta subcomplex, 4, 15kDa; NADH dehydrogenase (ubiquinone) 1 beta subcomplex, 5, 16kDa; NADH dehydrogenase (ubiquinone) 1 beta subcomplex, 6, 17kDa; NADH dehydrogenase (ubiquinone) 1 beta subcomplex, 7, 18kDa; NADH dehydrogenase (ubiquinone) 1 beta subcomplex, 8, 19kDa; NADH dehydrogenase (ubiquinone) 1 beta subcomplex, 9, 22kDa; NADH dehydrogenase (ubiquinone) 1, alpha/beta subcomplex, 1, 8kDa; NADH dehydrogenase (ubiquinone) 1, subcomplex unknown, 1 (6kD, KFYI); NADH dehydrogenase (ubiquinone) 1, subcomplex unknown, 1, 6kDa; NADH dehydrogenase (ubiquinone) 1, subcomplex unknown, 2, 14.5kDa; NADH dehydrogenase (ubiquinone) 42 kDa subunit; NADH dehydrogenase (ubiquinone) complex I, assembly factor 1; NADH dehydrogenase (ubiquinone) complex I, assembly factor 2; NADH dehydrogenase (ubiquinone) complex I, assembly factor 3; NADH dehydrogenase (ubiquinone) complex I, assembly factor 4; NADH dehydrogenase (ubiquinone) Fe-S protein 1, 75kDa (NADH-coenzyme Q reductase); NADH dehydrogenase (ubiquinone) Fe-S protein 2, 49kDa (NADH-coenzyme Q reductase); NADH dehydrogenase (ubiquinone) Fe-S protein 2-like (NADH-coenzyme Q reductase); NADH dehydrogenase (ubiquinone) Fe-S protein 3, 30kDa (NADH-coenzyme Q reductase); NADH dehydrogenase (ubiquinone) Fe-S protein 4 18kDa (NADH-coenzyme Q reductase) isoform NDUFS4_1; NADH dehydrogenase (ubiquinone) Fe-S protein 4 18kDa (NADH-coenzyme Q reductase) isoform NDUFS4_2; NADH dehydrogenase (ubiquinone) Fe-S protein 4, 18kDa (NADH-coenzyme Q reductase); NADH dehydrogenase (ubiquinone) Fe-S protein 5, 15kDa (NADH-coenzyme Q reductase); NADH dehydrogenase (ubiquinone) Fe-S protein 6, 13kDa (NADH-coenzyme Q reductase); NADH dehydrogenase (ubiquinone) Fe-S protein 7, 20kDa (NADH-coenzyme Q reductase); NADH dehydrogenase (ubiquinone) Fe-S protein 8, 23kDa (NADH-coenzyme Q reductase); NADH dehydrogenase (ubiquinone) flavoprotein 1, 51kDa; NADH dehydrogenase (ubiquinone) flavoprotein 2, 24kDa; NADH dehydrogenase (ubiquinone) flavoprotein 3, 10kDa; NADH dehydrogenase 1 alpha subcomplex assembly factor 2; NADH dehydrogenase 1 alpha subcomplex, 1; NADH dehydrogenase 1 beta subcomplex 8; NADH dehydrogenase [ubiquinone] 1 alpha subcomplex assembly factor 1; NADH dehydrogenase [ubiquinone] 1 alpha subcomplex assembly factor 2; NADH dehydrogenase [ubiquinone] 1 alpha subcomplex assembly factor 3; NADH dehydrogenase [ubiquinone] 1 alpha subcomplex assembly factor 4; NADH dehydrogenase [ubiquinone] 1 alpha subcomplex subunit 1; NADH dehydrogenase [ubiquinone] 1 alpha subcomplex subunit 10, mitochondrial; NADH dehydrogenase [ubiquinone] 1 alpha subcomplex subunit 11; NADH dehydrogenase [ubiquinone] 1 alpha subcomplex subunit 12; NADH dehydrogenase [ubiquinone] 1 alpha subcomplex subunit 13; NADH dehydrogenase [ubiquinone] 1 alpha subcomplex subunit 2; NADH dehydrogenase [ubiquinone] 1 alpha subcomplex subunit 3; NADH dehydrogenase [ubiquinone] 1 alpha subcomplex subunit 4; NADH dehydrogenase [ubiquinone] 1 alpha subcomplex subunit 4-like 2; NADH dehydrogenase [ubiquinone] 1 alpha subcomplex subunit 5; NADH dehydrogenase [ubiquinone] 1 alpha subcomplex subunit 6; NADH dehydrogenase [ubiquinone] 1 alpha subcomplex subunit 7; NADH dehydrogenase [ubiquinone] 1 alpha subcomplex subunit 8; NADH dehydrogenase [ubiquinone] 1 alpha subcomplex subunit 9, mitochondrial; NADH dehydrogenase [ubiquinone] 1 beta subcomplex subunit 1; NADH dehydrogenase [ubiquinone] 1 beta subcomplex subunit 10; NADH dehydrogenase [ubiquinone] 1 beta subcomplex subunit 11, mitochondrial; NADH dehydrogenase [ubiquinone] 1 beta subcomplex subunit 2, mitochondrial; NADH dehydrogenase [ubiquinone] 1 beta subcomplex subunit 3; NADH dehydrogenase [ubiquinone] 1 beta subcomplex subunit 4; NADH dehydrogenase [ubiquinone] 1 beta subcomplex subunit 5, mitochondrial; NADH dehydrogenase [ubiquinone] 1 beta subcomplex subunit 6; NADH dehydrogenase [ubiquinone] 1 beta subcomplex subunit 7; NADH dehydrogenase [ubiquinone] 1 beta subcomplex subunit 8, mitochondrial; NADH dehydrogenase [ubiquinone] 1 beta subcomplex subunit 9; NADH dehydrogenase [ubiquinone] 1 subunit C1, mitochondrial; NADH dehydrogenase [ubiquinone] 1 subunit C2; NADH dehydrogenase [ubiquinone] flavoprotein 1, mitochondrial; NADH dehydrogenase [ubiquinone] flavoprotein 2, mitochondrial; NADH dehydrogenase [ubiquinone] flavoprotein 3, mitochondrial; NADH dehydrogenase [ubiquinone] iron-sulfur protein 2, mitochondrial; NADH dehydrogenase [ubiquinone] iron-sulfur protein 3, mitochondrial; NADH dehydrogenase [ubiquinone] iron-sulfur protein 4, mitochondrial; NADH dehydrogenase [ubiquinone] iron-sulfur protein 5; NADH dehydrogenase [ubiquinone] iron-sulfur protein 6, mitochondrial; NADH dehydrogenase [ubiquinone] iron-sulfur protein 7, mitochondrial; NADH dehydrogenase [ubiquinone] iron-sulfur protein 8, mitochondrial; NADH dehydrogenase Fe-S protein 6; NADH dehydrogenase flavoprotein 1; NADH dehydrogenase flavoprotein 1; 24 kDa (FP); NADH dehydrogenase flavoprotein 2; NADH dehydrogenase subunit 1; NADH dehydrogenase subunit 3; NADH dehydrogenase subunit 4; NADH dehydrogenase subunit 6; NADH dehydrogenase subunit II; NADH dehydrogenase ubiquinone Fe-S 8; NADH dehydrogenase ubiquinone flavoprotein 1; NADH dehydrogenase ubiquinone flavoprotein 2, mitochondrial; NADH dehydrogenase-ubiquinone 30 kDa subunit; NADH ubiquinone oxidoreductase PDSW subunit (RH 16p13.3); NADH ubiquinone oxidoreductase subunit MWFE; NADH-coenzyme Q reductase; NADH-ubiquinone oxidoreductase 1 alpha subcomplex, 6; NADH-ubiquinone oxidoreductase 1 alpha subcomplex, assembly factor 1; NADH-ubiquinone oxidoreductase 13 kDa-A subunit; NADH-ubiquinone oxidoreductase 13 kDa-B subunit; NADH-ubiquinone oxidoreductase 15 kDa subunit; NADH-ubiquinone oxidoreductase 18 kDa subunit; NADH-ubiquinone oxidoreductase 19 kDa subunit; NADH-ubiquinone oxidoreductase 20 kDa subunit; NADH-ubiquinone oxidoreductase 23 kDa subunit; NADH-ubiquinone oxidoreductase 24 kDa subunit; NADH-ubiquinone oxidoreductase 30 kDa subunit; NADH-ubiquinone oxidoreductase 39 kDa subunit; NADH-ubiquinone oxidoreductase 42 kDa subunit; NADH-ubiquinone oxidoreductase 49 kDa subunit; NADH-ubiquinone oxidoreductase 51 kDa subunit; NADH-ubiquinone oxidoreductase 75 kDa subunit, mitochondrial; NADH-ubiquinone oxidoreductase 9 kD subunit; NADH-ubiquinone oxidoreductase 9 kDa subunit; NADH-ubiquinone oxidoreductase 9.6 kDa subunit; NADH-ubiquinone oxidoreductase AGGG subunit; NADH-ubiquinone oxidoreductase ASHI subunit; NADH-ubiquinone oxidoreductase B12 subunit; NADH-ubiquinone oxidoreductase B14 subunit; NADH-ubiquinone oxidoreductase B15 subunit; NADH-ubiquinone oxidoreductase B16.6 subunit; NADH-ubiquinone oxidoreductase B17 subunit; NADH-ubiquinone oxidoreductase B18 subunit; NADH-ubiquinone oxidoreductase B22 subunit; NADH-ubiquinone oxidoreductase B8 subunit; NADH-ubiquinone oxidoreductase B9 subunit; NADH-ubiquinone oxidoreductase beta subunit, 6; NADH-ubiquinone oxidoreductase chain 1; NADH-ubiquinone oxidoreductase chain 3; NADH-ubiquinone oxidoreductase chain 4; NADH-ubiquinone oxidoreductase chain 6; NADH-ubiquinone oxidoreductase complex; NADH-ubiquinone oxidoreductase ESSS subunit; NADH-ubiquinone oxidoreductase flavoprotein 2; NADH-ubiquinone oxidoreductase flavoprotein 3; NADH-ubiquinone oxidoreductase flavoprotein 3, 10kD; NADH-ubiquinone oxidoreductase KFYI subunit; NADH-ubiquinone oxidoreductase MLRQ subunit; NADH-ubiquinone oxidoreductase MLRQ subunit homolog; NADH-ubiquinone oxidoreductase MNLL subunit; NADH-ubiquinone oxidoreductase MWFE subunit; NADH-ubiquinone oxidoreductase NDUFS2 subunit; NADH-ubiquinone oxidoreductase PDSW subunit; NADH-ubiquinone oxidoreductase SGDH subunit; NADH-ubiquinone oxidoreductase subunit B14.5a; NADH-ubiquinone oxidoreductase subunit B14.5b; NADH-ubiquinone oxidoreductase subunit B14.7; NADH-ubiquinone oxidoreductase subunit B17.2; NADH-ubiquinone oxidoreductase subunit CI-B8; NADH-ubiquinone reductase 24 kDa mitochondrial; NADH:ubiquinone oxidoreductase (complex 1); NADH:ubiquinone oxidoreductase 15 kDa IP subunit; NADH:ubiquinone oxidoreductase ASHI subunit; NADH:ubiquinone oxidoreductase B15 subunit; NADH:ubiquinone oxidoreductase MLRQ subunit homolog; NADH:ubiquinone oxidoreductase NDUFS6 subunit; NADH:ubiquinone oxidoreductase PGIV subunit; NADH:ubiquinone oxidoreductase PSST subunit; NADH:ubiquinone oxidoreductase SDAP subunit; NADH:ubiquinone reductase precursor; NADHDH; NADHDH2; NDUFA12-like protein; NDUFA3; NDUFA4; NDUFA6; NDUFA7; NDUFB2; NDUFB3; NDUFB6; NDUFB7; NDUFB8; NDUFC1; NDUFC2; NDUFS5; NDUFS6; NDUFV1; NDUFV2; NDUFV3; Neuronal protein 15.6; Neuronal protein 17.3; Np15.6; Np17.3; nuclear protein 15.6; nuclear protein E3-3; nuclear-encoded mitochondrial NADH-ubiquinone reductase 24Kd subunit; NUOMS; p15.6; P17.3; Protein 2P1; Protein HRPAP20; PSST subunit; Renal carcinoma antigen NY-REN-4; short chain dehydrogenase/reductase family 22E, member 1; stuctural complex I protein; subunit of Mitochondrial Complex I; TYKY subunit; type I dehydrogenase; ubiquinone reductase; unnamed protein product; ZNF183

          View more View less

          Gene Aliases: 0610006N12Rik; 0610007D05Rik; 0610010M09Rik; 0610011B04Rik; 0610033L03Rik; 1010001M04Rik; 1010001M12Rik; 1010001N11Rik; 1110002H15Rik; 1110007M04Rik; 1190008J14Rik; 1300010H20Rik; 14.5kDa; 14kDa; 1500032D16Rik; 1700022J01Rik; 1810004I06Rik; 1810011O01Rik; 1810049F12Rik; 1810058I14Rik; 2010012C24Rik; 2010300P09Rik; 2210401F17Rik; 22kDa; 2310016K22Rik; 2310039H15Rik; 2400007M02Rik; 2410001M24Rik; 2410011G03Rik; 2610003B19Rik; 2700033I16Rik; 2700038D15Rik; 2700054G14Rik; 2900002J19Rik; 2900010C23Rik; 2900010I05Rik; 2900053E13Rik; 2P1; 3000003G13Rik; 4733401H18Rik; 5830412M15Rik; 6720411N02Rik; 8kDa; 9130423F15Rik; 9930026A05Rik; ACP; ACP1; AGGG; AI256693; AI325567; AI415450; AI987932; AL033311; AQDQ; ASHI; AU015782; AU022060; AV000592; AV006275; AW112974; AW214064; AW261656; B12; B13; B14; B14.5a; B14.5b; B14.7; B15; B16.6; B17; B17.2; B17.2L; B18; B22; B230217P19Rik; B8; B9; bA22L21.1; BC021616; BC059730; BC064011; C1-18k; C1-B8; C22H3orf60; C3orf60; C6orf66; C86051; C9H6ORF66; CC6; CD14; CDA016; CGI-39; CGI-65; CGI65; CI; CI-10k; CI-13kA; CI-13kB; CI-13kD-A; CI-13KD-B; CI-15k; CI-18; CI-18 kDa; CI-19KD; CI-20; CI-20KD; CI-23k; CI-23kD; CI-24k; CI-30; CI-30kD; CI-39k; CI-42k; CI-42KD; CI-49; CI-51K; CI-51kD; CI-75k; CI-75Kd; CI-9k; CI-9KD; CI-AGGG; CI-AQDQ; CI-ASHI; CI-B12; CI-B14; CI-B14-5A; CI-B14.5a; CI-B14.5b; CI-B14.7; CI-B15; CI-B18; CI-B22; CI-B8; CI-B9; CI-ESSS; CI-KFYI; CI-MLRQ; CI-MNLL; CI-MWFE; CI-PGIV; CI-SDAP; CI-SGDH; CI13KDA; CI15K; CI23KD; CI39k; CI51KD; CIA30; CIB8; CISGDH; COQ11; COXFA4; COXFA4L2; D5Bwg0566e; D5Bwg0577e; DAP13; E3-3; ESSS; FASN2A; G1; Gm137; GRIM-19; GRIM19; HLC-1; HLC1; HRPAP20; HSPC125; IP13; KFYI; LHONAR2; LYRM3; LYRM6; MC1DN1; MC1DN10; MC1DN11; MC1DN12; MC1DN13; MC1DN14; MC1DN15; MC1DN18; MC1DN2; MC1DN22; MC1DN23; MC1DN24; MC1DN25; MC1DN26; MC1DN28; MC1DN3; MC1DN30; MC1DN32; MC1DN33; MC1DN35; MC1DN36; MC1DN37; MC1DN39; MC1DN4; MC1DN5; MC1DN6; MC1DN7; MC1DN8; MC1DN9; MC4DN21; mimitin; MISTR1; MISTRH; MLRQ; MMTN; MNLL; MRCAF1; mt-Nd1; mt-Nd3; mt-Nd4; mt-Nd6; Mtnd1; Mtnd4; Mtnd6; MWFE; My013; MY017; NADHB14; NADHDH2; Nd1; Nd3; Nd4; Nd6; NDUFA1; NDUFA10; NDUFA11; NDUFA12; NDUFA12L; NDUFA13; NDUFA2; NDUFA3; NDUFA4; NDUFA4L2; NDUFA5; NDUFA6; NDUFA7; NDUFA8; NDUFA9; NDUFAB1; NDUFAF1; NDUFAF2; NDUFAF3; NDUFAF4; NDUFB1; NDUFB10; NDUFB11; NDUFB2; NDUFB3; NDUFB4; NDUFB5; NDUFB6; NDUFB7; NDUFB8; NDUFB9; NDUFC1; NDUFC2; NDUFS1; NDUFS2; NDUFS2L; NDUFS3; NDUFS4; NDUFS5; NDUFS6; NDUFS7; NDUFS8; NDUFV1; NDUFV2; NDUFV3; Np15; NP15.6; NP17.3; NUFM; NUOMS; P17.3; PDSW; PGIV; PRO1304; PSST; R75094; SDAP; SDR22E1; SGDH; TYKY; UNQ111/PRO1064; UQOR1; UQOR13; UQOR22; ZNF183

          View more View less

          UniProt ID: (Mouse) P03899, (Mouse) P03925, (Bovine) Q02377, (Human) O15239, (Human) O95299, (Bovine) P34942, (Bovine) Q8HXG6, (Human) Q86Y39, (Human) Q9UI09, (Bovine) O97725, (Bovine) Q95KV7, (Human) Q9P0J0, (Bovine) Q02370, (Human) O43678, (Bovine) Q02371, (Human) O95167, (Bovine) Q01321, (Human) O00483, (Bovine) Q3SZ44, (Human) Q9NRX3, (Bovine) P23935, (Human) Q16718, (Bovine) Q02366, (Human) P56556, (Bovine) Q05752, (Human) O95182, (Human) P51970, (Bovine) P42029, (Human) Q16795, (Bovine) P34943, (Human) O14561, (Bovine) P52505, (Human) Q9Y375, (Human) Q8N183, (Bovine) Q32P65, (Human) Q9BU61, (Bovine) Q2HJI2, (Human) Q9P032, (Bovine) A4FUH5, (Human) O75438, (Bovine) Q02378, (Bovine) Q02373, (Human) O96000, (Bovine) Q8HXG5, (Human) Q9NX14, (Bovine) Q02374, (Human) O95178, (Bovine) Q02365, (Human) O43676, (Human) O95168, (Bovine) P48305, (Human) O43674, (Bovine) Q02380, (Bovine) Q02367, (Human) O95139, (Bovine) Q02368, (Human) P17568, (Human) O95169, (Bovine) Q02372, (Human) Q9Y6M9, (Bovine) Q02369, (Human) O43677, (Bovine) Q02376, (Human) O95298, (Bovine) Q02827, (Bovine) P15690, (Human) P28331, (Bovine) P17694, (Human) O75306, (Bovine) P23709, (Human) O75489, (Bovine) Q02375, (Human) O43181, (Human) O43920, (Bovine) Q02379, (Human) O75380, (Bovine) P23934, (Human) O75251, (Bovine) P42026, (Human) O00217, (Bovine) P42028, (Human) P49821, (Bovine) P25708, (Bovine) P04394, (Human) P19404, (Human) P56181, (Bovine) P25712, (Mouse) O35683, (Mouse) Q99LC3, (Mouse) Q7TMF3, (Mouse) Q9ERS2, (Mouse) Q9CQ75, (Mouse) Q9CQ91, (Mouse) Q62425, (Mouse) Q4FZG9, (Mouse) Q9CPP6, (Mouse) Q9CQZ5, (Mouse) Q9Z1P6, (Mouse) Q9DCJ5, (Mouse) Q9DC69, (Mouse) Q9CR21, (Mouse) Q9CWX2, (Mouse) Q59J78, (Mouse) Q9JKL4, (Mouse) Q9D1H6, (Mouse) Q9DCS9, (Mouse) O09111, (Mouse) Q9CPU2, (Mouse) Q9CQZ6, (Mouse) Q9CQC7, (Mouse) Q9CQH3, (Mouse) Q3UIU2, (Mouse) Q9CR61, (Mouse) Q9D6J5, (Mouse) Q9CQJ8, (Mouse) Q9CQY9, (Mouse) Q9CQ54, (Mouse) Q91VD9, (Mouse) Q91WD5, (Mouse) Q9DCT2, (Mouse) Q9CXZ1, (Mouse) Q99LY9, (Mouse) P52503, (Mouse) Q9DC70, (Mouse) Q8K3J1, (Mouse) Q91YT0, (Mouse) Q9D6J6, (Mouse) Q8BK30

          View more View less

          Entrez Gene ID: (Mouse) 17718, (Mouse) 17722, (Bovine) 327673, (Human) 4694, (Human) 4705, (Bovine) 338060, (Bovine) 326346, (Human) 126328, (Human) 55967, (Bovine) 281742, (Bovine) 338084, (Human) 51079, (Bovine) 327698, (Human) 4695, (Bovine) 338064, (Human) 4696, (Bovine) 327704, (Human) 4697, (Bovine) 613541, (Human) 56901, (Bovine) 327714, (Human) 4698, (Bovine) 327670, (Human) 4700, (Bovine) 338063, (Human) 4701, (Human) 4702, (Bovine) 327710, (Human) 4704, (Bovine) 404188, (Human) 4706, (Bovine) 327702, (Bovine) 541232, (Human) 51103, (Human) 91942, (Bovine) 767961, (Human) 25915, (Bovine) 511968, (Human) 29078, (Bovine) 615440, (Human) 4707, (Bovine) 327690, (Bovine) 327701, (Human) 4716, (Bovine) 404161, (Human) 54539, (Bovine) 327713, (Human) 4708, (Bovine) 338073, (Human) 4709, (Human) 4710, (Bovine) 327706, (Human) 4711, (Bovine) 338061, (Bovine) 327665, (Human) 4712, (Bovine) 338065, (Human) 4713, (Human) 4714, (Bovine) 282517, (Human) 4715, (Bovine) 327660, (Human) 4717, (Bovine) 282289, (Human) 4718, (Bovine) 338046, (Bovine) 288380, (Human) 4719, (Bovine) 327697, (Human) 4720, (Bovine) 287327, (Human) 4722, (Bovine) 327680, (Human) 4724, (Human) 4725, (Bovine) 338057, (Human) 4726, (Bovine) 327691, (Human) 374291, (Bovine) 338079, (Human) 4728, (Bovine) 287027, (Human) 4723, (Bovine) 287014, (Bovine) 282290, (Human) 4729, (Human) 4731, (Bovine) 327717, (Mouse) 54405, (Mouse) 67273, (Mouse) 69875, (Mouse) 66414, (Mouse) 67184, (Mouse) 17991, (Mouse) 66091, (Mouse) 17992, (Mouse) 407790, (Mouse) 68202, (Mouse) 67130, (Mouse) 66416, (Mouse) 68375, (Mouse) 66108, (Mouse) 70316, (Mouse) 69702, (Mouse) 75597, (Mouse) 66706, (Mouse) 68493, (Mouse) 68342, (Mouse) 104130, (Mouse) 68198, (Mouse) 66495, (Mouse) 68194, (Mouse) 66046, (Mouse) 230075, (Mouse) 66916, (Mouse) 67264, (Mouse) 66218, (Mouse) 66377, (Mouse) 68197, (Mouse) 227197, (Mouse) 226646, (Mouse) 68349, (Mouse) 17993, (Mouse) 595136, (Mouse) 407785, (Mouse) 75406, (Mouse) 225887, (Mouse) 17995, (Mouse) 72900, (Mouse) 78330

          View more View less

          Function(s)
          NADH dehydrogenase (ubiquinone) activity protein binding electron carrier activity ATP binding endopeptidase activator activity cytochrome-c oxidase activity macromolecular complex binding structural constituent of ribosome oxidoreductase activity NADH dehydrogenase activity acyl binding ACP phosphopantetheine attachment site binding involved in fatty acid biosynthetic process structural molecule activity fatty acid binding calcium ion binding mitochondrial large ribosomal subunit binding unfolded protein binding calmodulin binding metal ion binding 2 iron, 2 sulfur cluster binding oxidoreductase activity, acting on NAD(P)H iron-sulfur cluster binding 4 iron, 4 sulfur cluster binding ubiquitin protein ligase binding quinone binding NAD binding oxygen sensor activity protease binding monooxygenase activity oxidoreductase activity, acting on NAD(P)H, quinone or similar compound as acceptor FMN binding RNA binding nucleoside kinase activity molecular_function protein complex binding poly(A) RNA binding oxidoreductase dehydrogenase metabolite interconversion enzyme transfer/carrier protein reductase nucleotide kinase chaperone
          Process(es)
          mitochondrial respiratory chain complex I assembly oxidation-reduction process mitochondrial electron transport, NADH to ubiquinone aerobic respiration mitochondrial ATP synthesis coupled proton transport hydrogen ion transmembrane transport mitochondrial ATP synthesis coupled electron transport response to oxidative stress electron transport chain negative regulation of cell growth cellular response to interferon-beta positive regulation of cysteine-type endopeptidase activity involved in apoptotic process protein import into mitochondrial inner membrane positive regulation of protein catabolic process negative regulation of transcription, DNA-templated cellular response to retinoic acid reactive oxygen species metabolic process apoptotic signaling pathway extrinsic apoptotic signaling pathway negative regulation of intrinsic apoptotic signaling pathway apoptotic process positive regulation of execution phase of apoptosis blastocyst hatching mitochondrial electron transport, cytochrome c to oxygen cellular respiration respiratory electron transport chain mitochondrial translation ATP synthesis coupled electron transport ubiquinone biosynthetic process sodium ion transport circadian rhythm lipid metabolic process fatty acid metabolic process fatty acid biosynthetic process protein lipoylation iron-sulfur cluster assembly [2Fe-2S] cluster assembly chaperone-mediated protein complex assembly protein-containing complex assembly cell projection organization cilium morphogenesis negative regulation of insulin secretion involved in cellular response to glucose stimulus NADH dehydrogenase complex assembly defense response to virus sensory perception of sound positive regulation of mitochondrial membrane potential negative regulation of NIK/NF-kappaB signaling negative regulation of reactive oxygen species biosynthetic process positive regulation of ATP biosynthetic process apoptotic mitochondrial changes ATP metabolic process regulation of mitochondrial membrane potential neurogenesis gliogenesis neural precursor cell proliferation cellular response to oxygen levels substantia nigra development regulation of protein phosphorylation brain development cAMP-mediated signaling positive regulation of fibroblast proliferation response to cAMP kidney development muscle contraction mitochondrion organization gene expression stem cell division DNA damage response, signal transduction by p53 class mediator multicellular organism growth reproductive system development circulatory system development mesenchymal stem cell differentiation cellular senescence mesenchymal stem cell proliferation electron transport coupled proton transport nervous system development cardiac muscle tissue development transport nucleobase-containing compound metabolic process biological_process ubiquinone-6 biosynthetic process positive regulation of cell proliferation negative regulation of apoptotic process mitophagy in response to mitochondrial depolarization multicellular organism aging mitochondrion morphogenesis cardiovascular system development
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-794c4b9cd6-hn5gs:80/100.66.79.229:80.
          git-commit: 7e8dd63fde7d4486b98ef9ac5cf4e365aa842ffe
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.52.0-Offline