Product Image
Applied Biosystems™

Path-ID™ qPCR Master Mix with Manual

Ambion™ Path-ID™ qPCR Master Mix is a convenient enzyme, buffer, and dNTP mix for TaqMan™ probe-based, real-time PCR detection ofRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNo. of Reactions
4388644M500 Reactions
4388643M100 Reactions
Catalog number 4388644M
Price (USD)
1,052.00
Each
No. of Reactions:
500 Reactions
Price (USD)
1,052.00
Each
Ambion™ Path-ID™ qPCR Master Mix is a convenient enzyme, buffer, and dNTP mix for TaqMan™ probe-based, real-time PCR detection of DNA targets. It is optimized for targets of importance in animal health and to function in the presence of PCR inhibitors. The mix is supplied in one tube suitable for 100 reactions. With the exception of PCR primers and probes for your target, the master mix contains everything you need for real-time PCR, including:
• AmpliTaq Gold™ ultrapure thermostable DNA polymerase designed for automatic hot start, to minimize nonspecific PCR products
• Optimized buffer and dNTP concentration, for reliable PCR
• Passive internal reference, based on proprietary ROX™ dye, for precise data analysis.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)7500 Fast System, 7500 System, 7900HT Fast System, 7900HT System,Applied Biosystems StepOnePlus™ Fast Real-Time PCR System, StepOne™, Standard Mode, StepOnePlus™, Standard Mode
FormatTube
Hot StartBuilt-In Hot Start
No. of Reactions500 Reactions
Passive Reference DyeROX (Pre-mixed)
PolymeraseAmpliTaq Gold DNA Polymerase
Product LineAmbion, Path-ID
Product TypeqPCR Master Mix
Purity or Quality GradeUP (Ultra Pure)
Quantity500 reactions
Sufficient For500 Reactions
Concentration2X
Detection MethodPrimer-probe
For Use With (Application)Animal Health, Pathogen Detection
FormLiquid
PCR MethodqPCR
Reaction SpeedFast or Standard
Unit SizeEach
Contents & Storage
2X qPCR Master Mix is stored at –20°C. Nuclease-free water may be stored at any temperature.