High-Capacity RNA-to-cDNA™ Kit, 500 reactions - FAQs
High-Capacity RNA-to-cDNA™ Kit, 500 reactions - FAQs
View additional product information for High-Capacity RNA-to-cDNA™ Kit - FAQs (4387406, 4388950)
3 product FAQs found
Which reverse transcription kit can I use with the PowerUp SYBR Green Master Mix (Cat. No. A25741, A25742, A25743, A25776, A25777, A25778, A25779, A25780, A25919)?
We recommend using the High-Capacity RNA-to-cDNA Kit (Cat. No. 4387406, 4388950).
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Applications Support Center.
Which kit should I use for the reverse transcription step for TaqMan Pri-miRNA assays (Cat. No. 4427012)?
We recommend the following kits:
  • High-Capacity cDNA Reverse Transcription Kit (Cat. No. 4374966).
  • High-Capacity RNA-to-cDNA Kit (Cat. No. 4387406).
Find additional tips, troubleshooting help, and resources within our PCR and cDNA Synthesis Support Center.
Is the 2X RT Buffer Mix within the High-Capacity RNA-to-cDNA Kit (Cat. N. 4387406, 4388950) available separately?
No, the 2X RT Buffer Mix is not available separately.
Find additional tips, troubleshooting help, and resources within our Nucleic Acid Purification and Analysis Support Center.