<i>Power</i> SYBR&trade; Green RNA-to-C<sub>T</sub>&trade; <i>1-Step</i> Kit
Applied Biosystems™

Power SYBR™ Green RNA-to-CT™ 1-Step Kit

The novel formulation reduces false positive results caused by primer dimers, significantly improves data accuracy, and provides reliable detection of low abundance targets.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNo. of Reactions
4391178100 Reactions
4389986200 Reactions
Catalog number 4391178
Price (USD)
251.65
Online exclusive
277.00
Save 25.35
Each
No. of Reactions:
100 Reactions
Price (USD)
251.65
Online exclusive
277.00
Save 25.35
Each

The novel formulation reduces false positive results caused by primer dimers, significantly improves data accuracy, and provides reliable detection of low abundance targets.

Get sensitivity and specificity in an easy-to-use one-step qRT-PCR reaction. The Power SYBR™ Green RNA-to-CT™ 1-Step kit has been formulated to ensure maximum sensitivity and reliability. The novel formulation reduces false positive results caused by primer dimers, significantly improves data accuracy, and provides reliable detection of low abundance targets. It includes:

  • AmpliTaq™ Gold DNA Polymerase UP for hot start qPCR, minimizing non-specific product formation
  • ArrayScript™ UP Reverse Transcriptase, an engineered RT that produces high yields of full-length cDNA
  • An additive that reduces primer dimer formation, reducing non-specific products that can cause false positive signals
  • RNase inhibitor, to prevent degradation of RNA templates
  • A proprietary ROX™ dye that serves as a passive internal reference to normalize non-PCR-related fluorescence fluctuations for superb precision on Applied Biosystems™ real-time PCR instruments.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)7500 Fast System, 7500 System, 7900HT System, QuantStudio™ 12k Flex, QuantStudio™ 3, QuantStudio™ 5, QuantStudio™ 6 Flex, QuantStudio™ 7, StepOne™, StepOnePlus™, ViiA™ 7 System
FormatTube
Kit TypeGreen RNA-to-CT 1-Step Kit
No. of Reactions100 Reactions
Passive Reference DyeROX (Pre-mixed)
PolymeraseAmpliTaq Gold DNA Polymerase
Product LineRNA-to-CT, SYBR
Product TypeReal Time PCR SYBR Master Mix
Purity or Quality GradeUP (Ultra Pure)
Quantity100 Reactions
Reverse TranscriptaseArrayScript™ UP
Run TimeStandard
Sample TypeRNA
Shipping ConditionWet Ice
Sufficient For100 Reactions at 20 μL
Volume1 x 1 mL
Concentration2X (Master Mix), 125X (RT Enzyme Mix)
Detection MethodSYBR
For Use With (Application)Gene Expression
Label or DyeSYBR Green
PCR Method1-step RT-qPCR
Reaction SpeedStandard
Unit SizeEach
Contents & Storage
Sufficient for 100 reactions at 20 μL reaction volume and includes:
• 1 x 1 mL tube of 2X master mix containing SYBR™Green 1 Dye, AmpliTaq Gold™ DNA Polymerase UP, dNTPs, Passive Reference 1, and optimized buffer components. Store at 2-8°C after opening.
• 1 x 20 μL tube of 125X RT enzyme mix containing ArrayScript™ UP Reverse Transcriptase and RNase Inhibitor. Store at -15 to -30°C.


Guaranteed minimum shelf life is 60 days (exact expiry date printed on product and CofA).

Frequently asked questions (FAQs)

What can I do to improve the sensitivity of my qPCR assay?

If you are targeting a low-abundance gene, you may have trouble getting Ct values in a good, reliable range (Ct > 32). To increase the sensitivity of the assay, you may want to consider the following:

- Increase the amount of RNA input into your reverse transcription reaction, if possible
- Increase the amount of cDNA in your qPCR reaction (20% by volume max)
- Try a different reverse transcription kit, such as our SuperScript VILO Master Mix, for the highest cDNA yield possible
- Consider trying a one-step or Cells-to-CT type workflow (depending on your sample type)

How do I set the baseline for my qPCR experiment?

Most times your instrument software can automatically set a proper baseline for your data. Check out our short video, Understanding Baselines, for more information on how to set them (https://www.youtube.com/watch?feature=player_embedded&v=5BjFAJHW-bE).

How do I set the threshold for my qPCR experiment?

In most cases your instrument software can automatically set a proper threshold for your data. Check out our short video, Understanding Thresholds, for more information on how to set them (https://www.youtube.com/watch?feature=player_embedded&v=H_xsuRQIM9M).

I am not getting any amplification with my TaqMan Assay or SYBR Green primer set. What is causing this?

There could be several reasons for no amplification from an assay or primer set. Please see these examples and suggested solutions in our Real-Time Troubleshooting Tool (https://www.thermofisher.com/us/en/home/life-science/pcr/real-time-pcr/qpcr-education/real-time-pcr-troubleshooting-tool/gene-expression-quantitation-troubleshooting/no-amplification.html) for more details.

I am getting amplification in my no-template control (NTC) wells in my qPCR experiment. What is causing this?

There could be several reasons for amplification in a NTC well. Please see these examples and suggested solutions in our Real-Time Troubleshooting Tool (https://www.thermofisher.com/us/en/home/life-science/pcr/real-time-pcr/qpcr-education/real-time-pcr-troubleshooting-tool/gene-expression-quantitation-troubleshooting/amplification-no-template-control.html) for more details.