TaqMan&trade; RNA-to-C<sub>T</sub>&trade; <i>1-Step</i> Kit
Actual product may vary
TaqMan&trade; RNA-to-C<sub>T</sub>&trade; <i>1-Step</i> Kit
TaqMan&trade; RNA-to-C<sub>T</sub>&trade; <i>1-Step</i> Kit
Applied Biosystems™

TaqMan™ RNA-to-CT™ 1-Step Kit

The Applied Biosystems TaqMan RNA-to-CT 1-Step Kit delivers consistent RNA target quantification for a wide variety of assays and isRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNo. of Reactions
43926562000 Reactions
439265340 Reactions
4392938200 Reactions
Catalog number 4392656
Price (USD)
9,240.65
Online exclusive
9,830.00
Save 589.35
Each
No. of Reactions:
2000 Reactions
Price (USD)
9,240.65
Online exclusive
9,830.00
Save 589.35
Each
The Applied Biosystems TaqMan RNA-to-CT 1-Step Kit delivers consistent RNA target quantification for a wide variety of assays and is validated with Applied Biosystems comprehensive library of TaqMan Gene Expression assays. The kit is recommended for a variety of applications including general gene expression studies, biomarker discovery, and microarray validation.

Features of the TaqMan RNA-to-CT 1-Step Kit include:
• Accurate across a wide dynamic range for dependable performance
• Validated with TaqMan Gene Expression Assays for easy experimental setup
• Consistent performance with a wide variety of targets, including AT-rich, GC-rich and long sequences
• 1-Step format means fewer pipetting steps leading to less chances for errors

Efficiency across a broad range of input concentrations
Maximum flexibility in RNA template input quantity requires optimal real-time PCR efficiency across a broad range of template quantities. Figure 1 shows the excellent amplification efficiency across six orders of magnitude. The result is comparable to that of the TaqMan RNA-to-CT 2-Step Kit, and is a significant improvement in dynamic range and sensitivity over the TaqMan One-Step RT-PCR Master Mix Reagents.

High sensitivity at low target concentration
The sensitivity of TaqMan RNA-to-CT 1-Step Kit was validated using a synthetic RNA template for which copy number is precisely known. Significant sampling error occurs when measuring low quantities of target, so statistical analysis is required for proper evaluation using multiple replicates. Figure 2 shows the expected quantity of target and corresponding mean CT values. Statistical analysis indicates high confidence of sample quantification based on a T-test (Table I), consistent with as few as 10 copies of target*.

*Results are dependent on a variety of factors, including assay design.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)7000 System, 7300 System, 7500 System, 7700 System, 7900HT System,Applied Biosystems StepOnePlus™ Fast Real-Time PCR System, StepOne™, Standard Mode, StepOnePlus™, Standard Mode
No. of Reactions2000 Reactions
Passive Reference DyeROX (Pre-mixed)
PolymeraseAmpliTaq Gold DNA Polymerase
Product LineRNA-to-CT, TaqMan
Product TypeOne-Step RT-PCR Kit
Purity or Quality GradeUP (Ultra Pure)
Quantity2,000 reactions
Reverse TranscriptaseArrayScript™ UP
Sample TypeRNA
Sufficient For2000 Reactions
Detection MethodPrimer-probe
For Use With (Application)Gene Expression
PCR Method1-step RT-qPCR
Reaction SpeedStandard
Unit SizeEach
Contents & Storage
Sufficient for 2000 reactions at 50 μL reaction volume. Includes:
• 10 x 5 mL tubes of 2X master mix containing AmpliTaq Gold™ DNA Polymerase UP, dNTPs (including dUTP), Passive Reference 1, and optimized buffer components. Store at -15 to -25°C.
• 10 x 250 μL tubes of 40X RT enzyme mix containing ArrayScript™ UP Reverse Transcriptase, RNase Inhibitor. Store at -15° to -30°C.

Guaranteed minimum shelf life is 60 days (exact expiry date printed on product and CofA).

Frequently asked questions (FAQs)

What can I do to improve the sensitivity of my qPCR assay?

If you are targeting a low-abundance gene, you may have trouble getting Ct values in a good, reliable range (Ct > 32). To increase the sensitivity of the assay, you may want to consider the following:

- Increase the amount of RNA input into your reverse transcription reaction, if possible
- Increase the amount of cDNA in your qPCR reaction (20% by volume max)
- Try a different reverse transcription kit, such as our SuperScript VILO Master Mix, for the highest cDNA yield possible
- Consider trying a one-step or Cells-to-CT type workflow (depending on your sample type)

How do I set the baseline for my qPCR experiment?

Most times your instrument software can automatically set a proper baseline for your data. Check out our short video, Understanding Baselines, for more information on how to set them (https://www.youtube.com/watch?feature=player_embedded&v=5BjFAJHW-bE).

How do I set the threshold for my qPCR experiment?

In most cases your instrument software can automatically set a proper threshold for your data. Check out our short video, Understanding Thresholds, for more information on how to set them (https://www.youtube.com/watch?feature=player_embedded&v=H_xsuRQIM9M).

I am not getting any amplification with my TaqMan Assay or SYBR Green primer set. What is causing this?

There could be several reasons for no amplification from an assay or primer set. Please see these examples and suggested solutions in our Real-Time Troubleshooting Tool (https://www.thermofisher.com/us/en/home/life-science/pcr/real-time-pcr/qpcr-education/real-time-pcr-troubleshooting-tool/gene-expression-quantitation-troubleshooting/no-amplification.html) for more details.

I am getting amplification in my no-template control (NTC) wells in my qPCR experiment. What is causing this?

There could be several reasons for amplification in a NTC well. Please see these examples and suggested solutions in our Real-Time Troubleshooting Tool (https://www.thermofisher.com/us/en/home/life-science/pcr/real-time-pcr/qpcr-education/real-time-pcr-troubleshooting-tool/gene-expression-quantitation-troubleshooting/amplification-no-template-control.html) for more details.