Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • ATP synthase Antibodies

          Invitrogen

          ATP synthase Monoclonal Antibody (12F4AD8AF8)

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite ATP synthase Monoclonal Antibody (12F4AD8AF8)

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (2)
          ATP synthase Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          ATP synthase Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 2

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          ATP synthase Antibody (43-9700) in ICC/IF

          Immunofluorescence analysis of ATP5A was performed using Hep G2 cells. The cells were fixed with 4% paraformaldehyde for 10 minutes, permeabilized with 0.1% Triton™ X-100 for 15 minutes, and blocked with 2% BSA for 1 hour at room temperature. The cells were labeled with ATP5A Mouse Monoclonal Antibody (Product # 43-9700) at 5 µg/mL in 0.1% BSA and incubated overnight at 4 degree and then labeled with Goat a... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          ATP synthase Antibody in Immunocytochemistry (ICC/IF)
          ATP synthase Antibody in Flow Cytometry (Flow)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Product Details

          43-9700

          Applications
          Tested Dilution
          Publications

          Immunocytochemistry (ICC/IF)

          5 µg/mL
          -

          Flow Cytometry (Flow)

          1 µg/ 10^6 cells
          -
          Product Specifications

          Species Reactivity

          Bovine, Human, Mouse

          Host/Isotype

          Mouse / IgG2b, kappa

          Class

          Monoclonal

          Type

          Antibody

          Clone

          12F4AD8AF8

          Immunogen

          Bovine Complex V

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Ammonium sulfate precipitation

          Storage buffer

          HEPES buffered saline

          Contains

          0.02% sodium azide

          Storage conditions

          4°C, do not freeze

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2533547

          Product Specific Information

          Positive control: Bovine heart mitochondria

          Target Information

          Complex V, also called F1F0ATPase or ATP synthase, is responsible for ATP production in oxidative phosphorylation and can work in reverse as a proton pumping ATPase. The enzyme was thought to be localized exclusively to mitochondria. However, it has recently been identified on the plasma membrane of several cell types including hepatocytes where it functions as the HDL receptor, on endothelial cells where it may act as the angiostatin receptor, and on the surface of cancer cells. The enzyme in mammals is composed of 17 subunits, five of which make up the easily detached F1. The remainder subunits are components of two stalk domains and the proton pumping F0 part of the machinery. Two of the subunits of the F0 part are encoded on mitochondrial DNA while the other subunits are nuclear encoded. Mutations in the mitochondrial-encoded subunits of ATP synthase (Complex V) cause OXPHOS disease.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: 6.8 kDa mitochondrial proteolipid protein; 6.8-kDa proteolipid with N-terminal sequence Met-Leu-Gln (MLQ); alpha subunit ATP synthase isoform precursor (EC 3.6.1.34); ATP synthase alpha chain, mitochondrial; ATP synthase alpha subunit; ATP synthase alpha subunit precursor; ATP synthase B chain, mitochondrial; ATP synthase c subunit; ATP synthase coupling factor VI; ATP synthase D chain, mitochondrial; ATP synthase e chain, mitochondrial; ATP synthase epsilon chain, mitochondrial; ATP synthase f chain, mitochondrial; ATP synthase F(0) complex subunit B1, mitochondrial; ATP synthase F1 subunit epsilon; ATP synthase g chain, mitochondrial; ATP synthase gamma chain, mitochondrial; ATP synthase gamma-subunit gene encoding a mitochondrial protein; ATP synthase inhibitor protein; ATP synthase lipid-binding protein, mitochondrial; ATP synthase membrane subunit 6.8PL; ATP synthase proteolipid P1; ATP synthase proteolipid P2; ATP synthase proteolipid P3; ATP synthase proton-transporting mitochondrial F(0) complex subunit B1; ATP synthase proton-transporting mitochondrial F(0) complex subunit C1; ATP synthase proton-transporting mitochondrial F(0) complex subunit C2; ATP synthase proton-transporting mitochondrial F(0) complex subunit C3; ATP synthase subunit 9; ATP synthase subunit ATP5MJ, mitochondrial; ATP synthase subunit ATP5MPL, mitochondrial; ATP synthase subunit b, mitochondrial; ATP synthase subunit beta, mitochondrial; ATP synthase subunit d, mitochondrial; ATP synthase subunit delta, mitochondrial; ATP synthase subunit e, mitochondrial; ATP synthase subunit epsilon, mitochondrial; ATP synthase subunit f, mitochondrial; ATP synthase subunit g, mitochondrial; ATP synthase subunit gamma, mitochondrial; ATP synthase subunit h; ATP synthase subunit O, mitochondrial; ATP synthase, H+ transporting mitochondrial F1 complex, alpha subunit; ATP synthase, H+ transporting mitochondrial F1 complex, beta subunit; ATP synthase, H+ transporting, mitochondrial F0 complex, subunit b; ATP synthase, H+ transporting, mitochondrial F0 complex, subunit B1; ATP synthase, H+ transporting, mitochondrial F0 complex, subunit c (subunit 9); ATP synthase, H+ transporting, mitochondrial F0 complex, subunit C1 (subunit 9); ATP synthase, H+ transporting, mitochondrial F0 complex, subunit C2 (subunit 9); ATP synthase, H+ transporting, mitochondrial F0 complex, subunit C3 (subunit 9); ATP synthase, H+ transporting, mitochondrial F0 complex, subunit D; ATP synthase, H+ transporting, mitochondrial F0 complex, subunit E; ATP synthase, H+ transporting, mitochondrial F0 complex, subunit F; ATP synthase, H+ transporting, mitochondrial F0 complex, subunit F2; ATP synthase, H+ transporting, mitochondrial F0 complex, subunit F6; ATP synthase, H+ transporting, mitochondrial F0 complex, subunit G; ATP synthase, H+ transporting, mitochondrial F1 complex, alpha subunit 1, cardiac muscle; ATP synthase, H+ transporting, mitochondrial F1 complex, beta polypeptide; ATP synthase, H+ transporting, mitochondrial F1 complex, delta subunit; ATP synthase, H+ transporting, mitochondrial F1 complex, epsilon subunit; ATP synthase, H+ transporting, mitochondrial F1 complex, gamma polypeptide 1; ATP synthase, H+ transporting, mitochondrial F1 complex, O subunit; ATP synthase, H+ transporting, mitochondrial F1 complex, O subunit; DNA segment, Chr 12, Wayne State University 28, expressed; ATP synthase, H+ transporting, mitochondrial F1F0 complex, subunit E; ATP synthase, H+ transporting, mitochondrial F1F0, subunit d; ATP synthase, H+ transporting, mitochondrial F1F0, subunit g; ATP synthase, H+ transporting, mitochondrial Fo complex subunit B1; ATP synthase, H+ transporting, mitochondrial Fo complex subunit C1 (subunit 9); ATP synthase, H+ transporting, mitochondrial Fo complex subunit C2 (subunit 9); ATP synthase, H+ transporting, mitochondrial Fo complex subunit C3 (subunit 9); ATP synthase, H+ transporting, mitochondrial Fo complex subunit D; ATP synthase, H+ transporting, mitochondrial Fo complex subunit E; ATP synthase, H+ transporting, mitochondrial Fo complex subunit F2; ATP synthase, H+ transporting, mitochondrial Fo complex subunit F6; ATP synthase, H+ transporting, mitochondrial Fo complex subunit G; ATP synthase, H+ transporting, mitochondrial Fo complex, subunit d; ATP synthase, mitochondrial, C subunit-3; ATP synthase-coupling factor 6, mitochondrial; ATP sythase (F1-ATPase) alpha subunit; Atp5b; ATP5C1; ATP5D; Atp5e; ATP5G1; ATP5G3; ATP5J; ATP5J2; ATP5L; ATP5O; ATPase inhibitor protein; ATPase inhibitory factor 1; ATPase protein 9; ATPase subunit 9; ATPase subunit b; ATPase subunit C; ATPase subunit d; ATPase subunit e; ATPase subunit epsilon; ATPase subunit F6; ATPase subunit G; ATPIF1; ATPIP; catalytic subunit; cell proliferation-inducing protein 47; coupling factor 6; dicyclohexylcarbodiimide (DCCD)-reactive proteolipid subunit; epididymis secretory protein Li 271; epididymis secretory sperm binding protein Li 123m; F(0)F(1)-ATPase; F-1-ATPase beta-subunit precursor; F-ATPase delta subunit; F-ATPase gamma subunit; F1 gamma; F1-ATPase beta-subunit; F1F0-ATP synthase g subunit; F1F0-ATP synthase, murine e subunit; F1F0-type ATP synthase subunit g; F1F0-type ATPase subunit f; F1Fo-ATP synthase complex Fo membrane domain f subunit; F1Fo-ATP synthase complex Fo membrane domain g subunit; F1Fo-ATPase synthase f subunit; Fog; H(+)-transporting two-sector ATPase; H+-ATP synthase subunit b; human ATP synthase OSCP subunit; hypothetical protein, clone:2-31; IF(1); IF1; inhibitor of F(1)F(o)-ATPase; mitochondrial ATP synthase beta subunit; mitochondrial ATP synthase complex delta-subunit precusor; mitochondrial ATP synthase epsilon chain; mitochondrial ATP synthase epsilon subunit; mitochondrial ATP synthase, coupling factor 6; mitochondrial ATP synthase, delta subunit; mitochondrial ATP synthase, gamma subunit 1; mitochondrial ATP synthase, H+ transporting F1 complex beta subunit; mitochondrial ATP synthase, subunit 9; mitochondrial ATP synthase, subunit C; mitochondrial ATP synthase, subunit F6; mitochondrial ATP synthetase, beta subunit; mitochondrial ATP synthetase, oligomycin-resistant; mitochondrial ATPase; mitochondrial ATPase coupling factor 6; My032 protein; oligomycin sensitivity conferral protein; oligomycin sensitivity conferral protein oscp-like protein; oligomycin sensitivity conferring protein; PIG36; PIG47; predicted protein of HQ1574; proliferation-inducing protein 36; proton-conducting channel, ATP synthase F(0) complex subunit c; unnamed protein product

          View more View less

          Gene Aliases: 0610008F14Rik; 0610009D10Rik; 1110019H14Rik; 1500000I11Rik; 1700094F02Rik; 2410043G19Rik; 2610008D24Rik; 4933437C06Rik; 6.8PL; APT5H; ATP5; ATP5A; ATP5A1; ATP5AL2; ATP5B; ATP5C; ATP5C1; ATP5CL1; ATP5D; ATP5E; ATP5F1; ATP5G; ATP5G1; ATP5G2; ATP5G3; ATP5H; ATP5I; ATP5J; ATP5J2; ATP5JG; ATP5JL; ATP5K; ATP5L; ATP5MPL; ATP5O; ATPE; ATPI; ATPIF1; ATPIP; ATPM; ATPMB; ATPO; ATPQ; ATPSB; C14orf2; CF6; COXPD22; D12Wsu28e; DYT38; DYTSPG; F6; hATP1; HEL-S-123m; HEL-S-271; HMC08D05; HUMOP2; IP; Lfm1; MC5DN3; MC5DN4; MC5DN4A; MC5DN4B; MC5DN5; MC5DN7; MLQ; MOM2; MP68; OMR; ORM; OSCP; P3; PIG47; PLPM

          View more View less

          UniProt ID: (Bovine) P19483, (Human) P25705, (Bovine) P00829, (Human) P06576, (Human) P36542, (Human) P30049, (Bovine) P05632, (Human) P56381, (Human) Q9UII2, (Human) P05496, (Human) Q06055, (Human) P48201, (Human) P56385, (Human) P56134, (Human) O75964, (Human) P56378, (Human) P24539, (Bovine) P13620, (Human) O75947, (Human) P18859, (Human) P48047, (Mouse) P00848, (Mouse) P56480, (Mouse) Q91VR2, (Mouse) Q9D3D9, (Mouse) P56382, (Mouse) Q9CR84, (Mouse) Q06185, (Mouse) P56135, (Mouse) Q9CPQ8, (Mouse) Q9CQQ7, (Mouse) Q9DCX2, (Mouse) P97450, (Mouse) Q9DB20

          View more View less

          Entrez Gene ID: (Bovine) 282578, (Human) 498, (Bovine) 327675, (Human) 506, (Human) 509, (Human) 513, (Bovine) 617230, (Human) 514, (Human) 93974, (Human) 516, (Human) 517, (Human) 518, (Human) 521, (Human) 9551, (Human) 10632, (Human) 9556, (Human) 515, (Bovine) 282710, (Human) 10476, (Human) 522, (Human) 539, (Mouse) 17705, (Mouse) 11947, (Mouse) 11949, (Mouse) 66043, (Mouse) 67126, (Mouse) 11951, (Mouse) 11958, (Mouse) 57423, (Mouse) 27425, (Mouse) 11950, (Mouse) 71679, (Mouse) 11957, (Mouse) 28080

          View more View less

          Function(s)
          protein binding protease binding RNA binding ATP binding ATPase activity adenyl ribonucleotide binding MHC class I protein binding ADP binding angiostatin binding proton-transporting ATP synthase activity, rotational mechanism proton-transporting ATPase activity, rotational mechanism structural molecule activity enzyme inhibitor activity calmodulin binding enzyme binding ATPase inhibitor activity identical protein binding ATPase binding mitochondrial proton-transporting ATP synthase complex binding hydrogen ion transmembrane transporter activity hydrogen ion channel activity macromolecular complex binding estradiol binding calcium ion binding lipoprotein particle receptor activity hydrogen-exporting ATPase activity, phosphorylative mechanism ligand-gated channel activity ATP synthase
          Process(es)
          negative regulation of endothelial cell proliferation lipid metabolic process ATP biosynthetic process response to muscle activity ATP synthesis coupled proton transport mitochondrial ATP synthesis coupled proton transport positive regulation of blood vessel endothelial cell migration response to ethanol ATP metabolic process cellular response to dexamethasone stimulus cellular response to nitric oxide hydrogen ion transmembrane transport angiogenesis osteoblast differentiation generation of precursor metabolites and energy regulation of intracellular pH oxidative phosphorylation aerobic respiration mitochondrial proton-transporting ATP synthase complex assembly response to copper ion response to ischemia heme biosynthetic process negative regulation of cardiac muscle cell apoptotic process erythrocyte differentiation positive regulation of protein catabolic process negative regulation of hydrolase activity mitochondrial depolarization regulation of protein localization to mitochondrion positive regulation of type 2 mitophagy negative regulation of mitochondrial ATP synthesis coupled proton transport substantia nigra development cellular response to cAMP cellular response to cytokine stimulus receptor-mediated endocytosis negative regulation of cell adhesion involved in substrate-bound cell migration cellular response to interleukin-7 response to peptide positive regulation of heart rate negative regulation of prostaglandin secretion positive regulation of blood pressure negative regulation of arachidonic acid secretion
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-76c4fffd86-nv6df:80/100.66.78.131:80.
          git-commit: bdc067a1e9e92d056899a2dda64af631988aa8a1
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.54.0-Offline